Merck
HPLC Analysis of Organic Acids on Discovery® C18
HPLC Analysis of Organic Acids on Discovery® C18 application for HPLC
Materiais
coluna analítica
Nº do produto
Descrição
Preços
Coluna de HPLC C18 Discovery®
5 μm particle size, L × I.D. 5 cm × 4.6 mm
CONDITIONS
column
Discovery C18, 5 cm × 4.6 mm I.D., 5 μm particles (504947)
mobile phase
[A] 0.1% TFA in [B] water:methanol; (60:40, A:B)
flow rate
2.0 mL/min
column temp.
20 °C
detector
UV, 254 nm