Merck
Methyl farnesoate plays a dual role in regulating Drosophila metamorphosis.
PLoS genetics (2015-03-17)
Di Wen, Crisalejandra Rivera-Perez, Mohamed Abdou, Qiangqiang Jia, Qianyu He, Xi Liu, Ola Zyaan, Jingjing Xu, William G Bendena, Stephen S Tobe, Fernando G Noriega, Subba R Palli, Jian Wang, Sheng Li
RESUMO
Corpus allatum (CA) ablation results in juvenile hormone (JH) deficiency and pupal lethality in Drosophila. The fly CA produces and releases three sesquiterpenoid hormones: JH III bisepoxide (JHB3), JH III, and methyl farnesoate (MF). In the whole body extracts, MF is the most abundant sesquiterpenoid, followed by JHB3 and JH III. Knockout of JH acid methyl transferase (jhamt) did not result in lethality; it decreased biosynthesis of JHB3, but MF biosynthesis was not affected. RNAi-mediated reduction of 3-hydroxy-3-methylglutaryl CoA reductase (hmgcr) expression in the CA decreased biosynthesis and titers of the three sesquiterpenoids, resulting in partial lethality. Reducing hmgcr expression in the CA of the jhamt mutant further decreased MF titer to a very low level, and caused complete lethality. JH III, JHB3, and MF function through Met and Gce, the two JH receptors, and induce expression of Kr-h1, a JH primary-response gene. As well, a portion of MF is converted to JHB3 in the hemolymph or peripheral tissues. Topical application of JHB3, JH III, or MF precluded lethality in JH-deficient animals, but not in the Met gce double mutant. Taken together, these experiments show that MF is produced by the larval CA and released into the hemolymph, from where it exerts its anti-metamorphic effects indirectly after conversion to JHB3, as well as acting as a hormone itself through the two JH receptors, Met and Gce.
MATERIAIS
Número do produto
Marca
Descrição do produto
Sigma-Aldrich
Diclorometano, suitable for HPLC, ≥99.8%, contains amylene as stabilizer
Sigma-Aldrich
Diclorometano, contains 40-150 ppm amylene as stabilizer, ACS reagent, ≥99.5%
Sigma-Aldrich
Diclorometano, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 50-150 ppm amylene as stabilizer
Sigma-Aldrich
3-Chloroperbenzoic acid, ≤77%
Sigma-Aldrich
Diclorometano, puriss. p.a., ACS reagent, reag. ISO, ≥99.9% (GC)
Sigma-Aldrich
Diclorometano, ACS reagent, ≥99.5%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Diclorometano, puriss., meets analytical specification of Ph. Eur., NF, ≥99% (GC)
Sigma-Aldrich
Diclorometano, ACS reagent, ≥99.5%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Diclorometano, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
DL-Methionine, ≥99%
Supelco
Diclorometano, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Diclorometano, biotech. grade, 99.9%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Juvenile hormone III, ≥65%, liquid, non-sterile
Supelco
Diclorometano, analytical standard
Sigma-Aldrich
DL-Methionine, 99%, FCC, FG
Sigma-Aldrich
DL-Methionine, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Supelco
Diclorometano, Selectophore, ≥99.5%
Sigma-Aldrich
Diclorometano, suitable for HPLC, ≥99.9%, contains 40-150 ppm amylene as stabilizer
L-Metionina, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
DL-Methionine, ≥99.0% (NT)
Supelco
Methoprene, PESTANAL®, analytical standard
DL-Methionine, European Pharmacopoeia (EP) Reference Standard