Documentos Principais
Z405051
Sigma-Aldrich
Aldrich® Sure/Stor flask
capacity 50 mL, plastic-coated glass
Selecione um tamanho
1 EA
R$ 2.911,00
R$ 2.911,00
Previsão de entrega em29 de maio de 2025
Selecione um tamanho
Alterar visualização
1 EA
R$ 2.911,00
About This Item
Código UNSPSC:
41121804
NACRES:
NB.43
R$ 2.911,00
Previsão de entrega em29 de maio de 2025
Materiais
plastic-coated glass
round bottom
three-neck
esterilidade
non-sterile
embalagem
pack of 1
fabricante/nome comercial
Sigma-Aldrich
capacidade
50 mL
Descrição geral
Designed for safe, reliable storage and dispensing of air-sensitive and odoriferous chemicals, pyrophorics, alkyl lithiums, Grignards, corrosives, and purified or deuterated solvents.
Features:
  • High-vacuum PTFE valve-to-glass seal eliminates air contamination in storage and septum leakage after initial needle penetration
  • Heavy-wall borosilicate glass
  • Removable side-arm hose connector for easy attachment of vacuum and inert gas lines. For long term storage, replace hose connector with a rubber septum for secondary protection. Flasks supplied with 2 septa. Order replacement septa, Z106496
  • Plastic-coated* flasks reduce chance of breakage and contain spills
*The coating is a thermoplastic film that softens appreciably at temperatures above 110°C, becoming less protective.Tests show that the coating is 100% effective in preventing flying glass when a fully evacuated flask is broken.
Sure/Stor Technique
1.
Connect flask to vacuum line to pump out air and strong odor, then refill with inert gas
2. Open PTFE valve sufficiently to allow syringe needle (or pipette) to pass through septum-inlet and reach the stored liquid in flask
3. Close the PTFE valve after needle removal and pump out again to remove air and odor (use rubber septa Z553913 or Z554030 on septum-inlet)
4. For long-term storage, replace hose connector with a rubber septum for secondary protection
NOTICE: We supply high quality PTFE valves on Aldrich brand glassware that has been tested and approved by our Glass Center.
Informações legais
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Sure/Stor is a trademark of Sigma-Aldrich Co. LLC
Escolha uma das versões mais recentes:
Certificados de análise (COA)
Visite a Biblioteca de Documentos
Conteúdo relacionado
Packaging options for solvents include glass, PVC coated, Sure/Seal bottles, and aluminum cans, offering diverse dispensing solutions.