TEM-CF150-CU-ET
Carbon Film Supported Copper Square Mesh
Extra Thick, size 150 mesh, box of 50
embalagem
box of 50
tamanho
150 mesh
Categorias relacionadas
Descrição geral
A 20-30 nm thick film of pure carbon supported standard squre mesh grids. Copper grid, 150 mesh.