27150-U
Vials, screw top with solid green Thermoset cap with PTFE liner, preassembled, pkg of 100
volume 7 mL, clear glass vial, O.D. × H 17 mm × 60 mm, thread for 15-425
Synonym(s):
PK100 7ML CLEAR VIAL,SCREW TOP,SOLID CAP W/PTFE LINER
About This Item
Recommended Products
material
F217/PTFE cap liner
PTFE liner
clear glass vial
green melamine resin cap (solid-top)
feature
closure type green melamine solid-top cap
closure type screw top vial
packaging
pkg of 100 ea
O.D. × H
17 mm × 60 mm
volume
7 mL
fitting
thread for 15-425
Related Categories
General description
related product
Choose from one of the most recent versions:
Certificates of Analysis (COA)