Skip to Content
Merck
All Photos(1)

Key Documents

27152

Supelco

Screw cap, solid top with PTFE liner, pkg 100

green melamine resin solid cap, F217/PTFE liner, for use with 7 mL vial with 15-425 thread

Sign Into View Organizational & Contract Pricing

Select a Size

100 EA
$130.00

$130.00


Available to ship on14 April 2025Details


Request a Bulk Order

Select a Size

Change View
100 EA
$130.00

About This Item

UNSPSC Code:
41122001

$130.00


Available to ship on14 April 2025Details


Request a Bulk Order

material

F217/PTFE liner
green melamine resin solid cap

size

15-425

feature

closure type screw thread, green melamine solid cap, PTFE liner

packaging

pkg of 100 ea

parameter

49 °C temp. limit

fitting

thread for 15-425

compatibility

for use with 7 mL vial with 15-425 thread

Looking for similar products? Visit Product Comparison Guide

General description

Green Thermoset Plastic Caps with F-217 PTFE Liners are known for providing the widest range of chemical compatibility and for being the most temperature tolerant of plastic closures. The color green easily identifies the PTFE liner system. F-217 foam-backed PTFE liners resist attack from virtually all chemicals at room temperature and provides odor and impact resilience for a tight seal with a low moisture transmission rate. F-217 has a low density foam core between two solid layers of LDPE (low density polyethylene). Some uses for these F-217/PTFE lined caps include analytical lab samples, high purity chemicals, strong acids solvents, environmental samples, pharmaceuticals, and diagnostic reagents.

Application

Amber vials fitted with PTFE-lined screw caps may be used to store soil sample extracts at -20 °C under nitrogen, in an experimental study demonstrating usage of lignin-degrading fungi for bioaugmentation, as a viable solid-phase treatment option, for the remediation of pentachlorophenol (PCP) contaminated-soils.[1]

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Solid-phase treatment of a pentachlorophenol-contaminated soil using lignin-degrading fungi.
Lamar RT
Environmental Science & Technology, 27 (12), 2566-2571 (1993)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service