Skip to Content
Merck
All Photos(1)

Key Documents

27138

Supelco

Vials, screw top with solid green Thermoset cap with PTFE liner, preassembled, pkg of 100

volume 4 mL, clear glass vial, O.D. × H 15 mm × 45 mm, thread for 13-425

Synonym(s):

PK100 4ML CLEAR VIAL,SCREW TOP,SOLID CAP W/PTFE LINER

Sign Into View Organizational & Contract Pricing

Select a Size

100 EA
$224.00

$224.00


Available to ship on22 April 2025Details


Request a Bulk Order

Select a Size

Change View
100 EA
$224.00

About This Item

UNSPSC Code:
41122001

$224.00


Available to ship on22 April 2025Details


Request a Bulk Order

material

F217/PTFE cap liner
PTFE liner
clear glass vial
green melamine resin cap (solid-top)

feature

closure type green melamine solid-top cap
closure type screw top vial

packaging

pkg of 100 ea

O.D. × H

15 mm × 45 mm

volume

4 mL

fitting

thread for 13-425

Looking for similar products? Visit Product Comparison Guide

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
271812718227000
volume

4 mL

volume

40 mL

volume

40 mL

volume

2 mL

fitting

thread for 13-425

fitting

thread for 24-400

fitting

thread for 24-400

fitting

thread for 8-425

material

F217/PTFE cap liner, clear glass vial, PTFE liner, green melamine resin cap (solid-top)

material

F217/PTFE cap liner, PTFE liner, clear glass vial, green melamine resin cap (solid-top)

material

F217/PTFE cap liner, PTFE liner, amber glass vial, green melamine resin cap (solid-top)

material

F217/PTFE cap liner, PTFE liner, amber glass vial (standard opening - 4.6 mm), green melamine resin cap (solid-top)

packaging

pkg of 100 ea

packaging

pkg of 100 ea

packaging

pkg of 100 ea

packaging

pkg of 100 ea

O.D. × H

15 mm × 45 mm

O.D. × H

29 mm × 82 mm

O.D. × H

29 mm × 82 mm

O.D. × H

-

Application

It has been used in decanting serum for residue analysis using GC/MS/MS.[1] It has also been used as reaction and development chamber in examining reaction of dioxane solution of oxalyl chloride with an electropositive metal such as Zn and Mg.[2]

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zn-Mediated Reduction of Oxalyl Chloride Forming CO and Its Application in Carbonylation Reactions.
Markovic?, Martin, et al.
Organic Letters, 17.22, 5618-5621 (2015)
Human maternal and umbilical cord blood concentrations of polybrominated diphenyl ethers.
Foster, Warren G., et al.
Chemosphere, 84.10, 1301-1309 (2011)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service