Merck
V800332
Sigma-Aldrich
Kaliumpermanganat
LR, ≥99%
About This Item
Empirische Formel (Hill-System):
KMnO4
CAS-Nummer:
Molekulargewicht:
158.03
EG-Nummer:
MDL-Nummer:
UNSPSC-Code:
12352312
PubChem Substanz-ID:
Qualität
LR
Produktlinie
Vetec
Assay
≥99%
Form
crystals
Anionenspuren
chloride (Cl-): ≤0.01%
sulfate (SO42-): ≤0.03%
SMILES String
[K+].[O-][Mn](=O)(=O)=O
InChI
1S/K.Mn.4O/q+1;;;;;-1
InChIKey
VZJVWSHVAAUDKD-UHFFFAOYSA-N
Anwendung
Geeignet für heterogene Oxidationen
Rechtliche Hinweise
Vetec is a trademark of Merck KGaA, Darmstadt, Germany
Signalwort
Danger
Gefahreneinstufungen
Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Ox. Sol. 2 - Repr. 2 - Skin Corr. 1 - STOT RE 2 Inhalation
Zielorgane
Brain
Lagerklassenschlüssel
5.1B - Oxidizing hazardous materials
WGK
WGK 3
Flammpunkt (°F)
Not applicable
Flammpunkt (°C)
Not applicable
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Die Dokumentenbibliothek aufrufen
Elisangela P Da Silva et al.
Journal of colloid and interface science, 555, 373-382 (2019-08-10)
In recent years, electrochemical energy devices, i.e. batteries, fuel cells, solar cells, and supercapacitors, have attracted considerable attention of scientific community. The architecture of active materials plays a crucial role for improving supercapacitors performance. Herein, titanium dioxide (TiO2) nanofibers (1D)
Jenny Li-Ying Huang et al.
Cancer research, 75(16), 3384-3397 (2015-06-27)
GNAQ and GNA11 are heterotrimeric G protein alpha subunits, which are mutated in a mutually exclusive pattern in most cases of uveal melanoma, one of the most aggressive cancers. Here we introduce the first transgenic mouse model of uveal melanoma
F C Holstege et al.
Methods (San Diego, Calif.), 12(3), 203-211 (1997-07-01)
Open complex formation precedes initiation of transcription by RNA polymerases. In the analysis of transcription initiation from eukaryotic class II promoters, we have used promoter DNA structures that represent intermediates in open complex formation. We describe the preparation and isolation
Ming Yang et al.
Nature communications, 6, 6445-6445 (2015-03-12)
Clostrubin is a potent antibiotic against methicillin- and vancomycin-resistant bacteria that was isolated from a strictly anaerobic bacterium Clostridium beijerinckii in 2014. This polyphenol possesses a fully substituted arene moiety on its pentacyclic scaffold, which poses a considerable challenge for
Zhanchao Meng et al.
Nature communications, 6, 6096-6096 (2015-02-05)
Indolosesquiterpenoids are a growing class of natural products that exhibit a wide range of biological activities. Here, we report the total syntheses of xiamycin A and oridamycins A and B, indolosesquiterpenoids isolated from Streptomyces. Two parallel strategies were exploited to