Wichtige Dokumente
22349
Diagrammpapier für Schreiber
for use with Linear Instruments 1200, 1201, 1202, 1210, 2020, 2030, chart type, 0100-0026, pkg of 25 ea
Alle Fotos(1)
About This Item
UNSPSC-Code:
14111500
Empfohlene Produkte
Leistungsmerkmale
chart type 0100-0026
Verpackung
pkg of 25 ea
Kompatibilität
for use with Linear Instruments 1200, 1201, 1202, 1210, 2020, 2030
Anwendung
Analog signal from the chromatograph may be connected to a recorder with chart paper speed of 5 min/in. during an analysis to measure Glutethimide concentrations in the serum and cerebrospinal fluid (CSF) using gas chromatography.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Comparison of glutethimide concentration in the serum and cerebrospinal fluid of humans in drug overdose.
M Gold et al.
Clinical chemistry, 19(10), 1158-1161 (1973-10-01)
A.V.Bakshi U.A.Bakshi
Electronic Measurements & Instrumentation (2008)
John Kenkel
Analytical Chemistry for Technicians , 366-366 (1994)