Merck
Z710210
Nichipet Eco pipette
volume 1000-10,000 μL
Größe auswählen
1 EA
€ 676,00
€ 676,00
Check Cart for Availability
Größe auswählen
Ansicht ändern
1 EA
€ 676,00
About This Item
UNSPSC-Code:
41121500
NACRES:
NB.01
€ 676,00
Check Cart for Availability
Beschreibung
With glass tip
Leistungsmerkmale
electronic: no
single-channel
Verpackung
pack of 1 ea
Hersteller/Markenname
Nichiryo 00-NPC-10ML
Messbereich Volumen
1000-10,000 μL
Volumen
10000 μL
Verwandte Kategorien
Allgemeine Beschreibung
Providing the accuracy and precision of a micropipette with the compatibility of glass. Glass tips allow pipetting of substances not compatibile with plastic.
  • Glass tip enables totally bias free liquid handling for endocrine disruptors analysis
  • 4 sizes cover a wide volume range (20-10000 μL)
  • Nozzle with double O-rings enables easy tip fitting/ejection
  • Reliable sealing nozzle is PTFE and piston is ceramic
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)