Größe auswählen
| Ihnen/SKU | Verfügbarkeit | Preis |
|---|---|---|
1 ea | Warenkorb auf Verfügbarkeit prüfen | € 637,00 |
Über diesen Artikel
€ 637,00
description
With glass tip
feature
electronic: no, single-channel
packaging
pack of 1 ea
manufacturer/tradename
Nichiryo 00-NPC-1000
volume measuring range
100-1000 μL
volume
1000 μL
General description
- Glass tip enables totally bias free liquid handling for endocrine disruptors analysis
- 4 sizes cover a wide volume range (20-10000 μL)
- Nozzle with double O-rings enables easy tip fitting/ejection
- Reliable sealing nozzle is PTFE and piston is ceramic
1 of 1
Dieser Artikel | |||
|---|---|---|---|
| description volume 100-1000 μL | description volume 1000-10,000 μL | description volume 20-200 μL | description volume 1000-5000 μL |
| manufacturer/tradename Nichiryo 00-NPC-1000 | manufacturer/tradename Nichiryo 00-NPC-10ML | manufacturer/tradename Nichiryo 00-NPC-200 | manufacturer/tradename Nichiryo 00-NPC-5000 |
| volume 1000 μL | volume 10000 μL | volume 200 μL | volume 5000 μL |
| feature single-channel, electronic: no | feature single-channel, electronic: no | feature electronic: no, single-channel | feature single-channel, electronic: no |
| packaging pack of 1 ea | packaging pack of 1 ea | packaging pack of 1 ea | packaging pack of 1 ea |
| volume measuring range 100-1000 μL | volume measuring range 1000-10,000 μL | volume measuring range 20-200 μL | volume measuring range 1000-5000 μL |
| description With glass tip | description With glass tip | description With glass tip | description With glass tip |
Still not finding the right product?
Explore all of our products under Nichipet Eco pipette
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?