Merck
Z687731
Replacement NMR needle with bevel tip for eVol® electronic syringe
for 500 μL, L × O.D. 11.5 cm × 0.63 mm, gauge size 23
Größe auswählen
5 EA
€ 111,00
€ 111,00
Voraussichtliches Versanddatum06. April 2025
Größe auswählen
Ansicht ändern
5 EA
€ 111,00
About This Item
UNSPSC-Code:
41121705
NACRES:
SB.32
€ 111,00
Voraussichtliches Versanddatum06. April 2025
Manometergröße
23
Hersteller/Markenname
SGE 039160
L × AD
11.5 cm × 0.63 mm
Allgemeine Beschreibung
The eVol® NMR electronic syringe facilitates in-tube sample mixing and enables recovery of precious samples allowing them to be stored in alternative vessels (such as inexpensive vials) rather than being left in valuable tubes. The beveled tip replacement needles come in multiple length and gauge options to fit your specific needs.
Rechtliche Hinweise
eVol is a registered trademark of SGE Analytical Science Pty Ltd.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)