Merck
BR782301
BRAND® strip plates
size 96 wells, 12xF8, MB, dividable: yes
Synonym(e):
BRANDplates®, Micro Tubes with Screw caps, Microcentrifuge Tubes, Microplates, Plates, Screw caps, Strip Plates
Größe auswählen
100 EA
€ 669,00
€ 669,00
Check Cart for Availability
Größe auswählen
Ansicht ändern
100 EA
€ 669,00
About This Item
UNSPSC-Code:
41121800
NACRES:
NB.11
€ 669,00
Check Cart for Availability
Materialien
polystyrene
transparent wells
Sterilität
non-sterile
Leistungsmerkmale
12xF8, MB
pureGrade
Verpackung
pack of 100 ea
pkg of (4 bags of 25)
Hersteller/Markenname
BRAND 782301
Größe
96 wells
Bindungsart
medium binding surface
Allgemeine Beschreibung
  • BRAND® strip plates are polystyrene, transparent with grid, strips of 8 wells, divisible and flat bottom.
  • Optimal surfaces enable optimal results.
  • Reliable quality and tested by independent laboratories.
  • Ideal visual properties and excellent flexibility.
  • In accordance with ANSI/SLAS production standards.
Leistungsmerkmale und Vorteile
  • Optimal surfaces for optimal results
  • Reliable quality from the cleanroom
  • Tested by independent laboratories
Rechtliche Hinweise
BRAND is a registered trademark of BRAND GMBH + CO KG
BRANDplates is a registered trademark of BRAND GMBH + CO KG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)