Größe auswählen
| Ihnen/SKU | Verfügbarkeit | Preis |
|---|
Über diesen Artikel
material
flat bottom clear polystyrene (96-well), round wells
sterility
non-sterile
feature
lid: no
packaging
plate of 12 strips
manufacturer/tradename
Corning 2580
size
96 wells
well volume
360 μL
well working volume
75-200 μL , for low volume plates volume is 50-125 μL
color
clear
suitability
suitable for (assay)
binding type
high binding surface
General description
Legal Information
1 of 1
Dieser Artikel | |||
|---|---|---|---|
| description high binding surface, without stripwell holder | description medium binding surface, clear | description low volume, high binding surface, clear | description low volume, medium binding surface, black |
| size 96 wells | size 96 wells | size 96 wells | size 96 wells |
| suitability suitable for (assay) | suitability suitable for (assay) | suitability suitable for (assay) | suitability suitable for (assay) |
| material flat bottom clear polystyrene (96-well), round wells | material flat bottom clear polystyrene (96-well), round wells | material clear polystyrene plate, flat bottom clear polystyrene (96-well), round clear wells | material black polystyrene plate, flat bottom clear polystyrene (96-well), round black wells |
| well volume 360 μL | well volume 360 μL | well volume 190 μL | well volume 190 μL |
| binding type high binding surface | binding type non-treated surface | binding type high binding surface | binding type non-treated surface |
| feature lid: no | feature lid: no, skirt | feature lid: no, skirt | feature lid: no, skirt |
Still not finding the right product?
Explore all of our products under Corning® 1 x 8 Stripwell™ 96-Well-Platten
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?