Merck
BR476000
BRAND® slide box
Holds 100 x slides 76x26 mm
Synonym(e):
Microscope slides, Microscopy, Slide box, Staining, Staining Accessories
Größe auswählen
1 EA
€ 38,20
€ 38,20
Check Cart for Availability
Größe auswählen
Ansicht ändern
1 EA
€ 38,20
About This Item
UNSPSC-Code:
41123302
NACRES:
MA.01
€ 38,20
Check Cart for Availability
Materialien
polystyrene
Leistungsmerkmale
Holds 100 x slides 76x26 mm
Verpackung
pack of 1 ea
Hersteller/Markenname
BRAND 476000
Allgemeine Beschreibung
Sturdy design. Screw lid with dealing wire lug. Ideal for storing or shipping 5 thick or 10 thin slides. Slides protrude 10 mm above the box, to facilitate removal.
For slides, size (in, mm) 76 x 26. Numbered slots in base. Lid with index card.
Rechtliche Hinweise
BRAND is a registered trademark of BRAND GMBH + CO KG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)