Merck
913839
Sigma-Aldrich
2-Methylnicotinic acid imidazolide
Synonym(e):
(1H-imidazol-1-yl)(2-methylpyridin-3-yl)methanone, 1H-Imidazol-1-yl(2-methyl-3-pyridinyl)methanone, 3-(1H-imidazole-1-carbonyl)-2-methylpyridine, NAI, RNA SHAPE probe, RNA structure probe, mNAI
Größe auswählen
50 MG
€ 85,70
500 MG
€ 267,00
€ 85,70
Check Cart for Availability
Bulk-Bestellung anfordern
Größe auswählen
Ansicht ändern
50 MG
€ 85,70
500 MG
€ 267,00
About This Item
Empirische Formel (Hill-System):
C10H9N3O
CAS-Nummer:
Molekulargewicht:
187.20
MDL-Nummer:
UNSPSC-Code:
12352106
NACRES:
NA.22
€ 85,70
Check Cart for Availability
Bulk-Bestellung anfordern
Form
powder
Qualitätsniveau
mp (Schmelzpunkt)
68-72 °C
Lagertemp.
2-8°C
SMILES String
[n]2(cncc2)C(=O)c1c(nccc1)C
InChI
1S/C10H9N3O/c1-8-9(3-2-4-12-8)10(14)13-6-5-11-7-13/h2-7H,1H3
InChIKey
OWQPEDNXDCVXJO-UHFFFAOYSA-N
Anwendung
2-Methylnicotinic acid imidazolide (NAI or mNAI) is an RNA SHAPE probe for live-cell RNA structure profiling. SHAPE -- or selective 2′-hydroxyl acylation analyzed by primer extension -- uses small, electrophilic chemical probes to react with the 2′-hydroxyl group and provides insight to RNA structure and its dynamics in varied cells states or in cells with gene knockouts. It has also been extended to the SHALiPE approach, selective 2′-hydroxyl acylation with lithium ion-based primer extension.
Ähnliches Produkt
Produkt-Nr.
Beschreibung
Preisangaben
Lagerklassenschlüssel
11 - Combustible Solids
WGK
WGK 3
Flammpunkt (°F)
Not applicable
Flammpunkt (°C)
Not applicable
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Die Dokumentenbibliothek aufrufen
Robert C Spitale et al.
Nature chemical biology, 9(1), 18-20 (2012-11-28)
RNA structure has important roles in practically every facet of gene regulation, but the paucity of in vivo structural probes limits current understanding. Here we design, synthesize and demonstrate two new chemical probes that enable selective 2'-hydroxyl acylation analyzed by
Shao-Ru Wang et al.
Nature communications, 11(1), 91-91 (2020-01-05)
Prokaryotes use repetitive genomic elements termed CRISPR (clustered regularly interspaced short palindromic repeats) to destroy invading genetic molecules. Although CRISPR systems have been widely used in DNA and RNA technology, certain adverse effects do occur. For example, constitutively active CRISPR