Skip to Content
MilliporeSigma
All Photos(1)

Documents

NIST1566B

Oyster tissue

NIST® SRM® 1566b

Synonym(s):

Oyster Sample

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

packaging

pkg of 25 g

manufacturer/tradename

NIST®

format

matrix material

General description

This Standard Reference Material (SRM) is intended primarily for use in evaluating analytical methods and instruments used for the determination of the mass fraction values of selected elements and proximates in marine bivalve tissue, foods, or similar materials. A unit of SRM 1566b contains approximately 25 g of freeze-dried oyster tissue. For more information, please refer to the COA and SDS.

SRM 1566B_cert

SRM 1566B_SDS

Application


  • Seafood Safety Analysis: Oyster tissue is critical in environmental and food safety analysis, especially for the determination of technology-critical elements using advanced spectrometry techniques, underpinning its importance in shellfish biochemistry and marine organism sample analysis (Wippermann et al., 2024).

Other Notes

Certified for the analytes listed below. See certificate for values and more details
Ash, Total Dietary Fiber, Moisture, Nitrogen (N), Protein Nitrogen, Protein, Solids
Trace Elements: Aluminum (Al), Antimony (SB), Arsenic (As), Barium (Ba), Boron (B), Cadmium (Cd), Calcium (Ca), Chlorine (Cl2), Cobalt (Co), Copper (Cu), Gold (Au), Hydrogen (H), Iron (Fe), Lead (Pb), Magnesium (Mg), Manganese (Mn), Mercury (Hg), Methylmercury (as mercury), Nickel (Ni), Potassium (K), Rubidium (Rb), Selenium (Se), Silver (Ag), Sodium (Na), Strontium (Sr), Sulfur (S), Thorium (Th), Tin (Sn), Titanium (Ti), Uranium (U), Vanadium (V), Zinc (Zn)

Matrix Group: Fish and Seafood

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service