Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

NIST3234

Soy flour

NIST® SRM® 3234

Synonym(s):

Soybean flour

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

packaging

pkg of 50 g

manufacturer/tradename

NIST®

format

matrix material

Looking for similar products? Visit Product Comparison Guide

General description

Each unit of SRM 3234 comprises a single bottle containing approximately 50 g of defatted soy flour, sealed in an aluminized pouch. For more information, please refer to the COA and SDS.
SRM 3234_cert

SRM 3234_SDS

Application

The certified values provided by this Standard Reference Material (SRM) are intended for validating methods used to determine elements in soy flour and similar materials, and they can also be utilized for quality assurance, such as when assigning values to in-house control materials.

Features and Benefits

  • Certified values for elements are reported on a dry-mass basis and traceable to SI derived unit of mass fraction and expressed as milligrams per kilogram.
  • Accompanied by non-certified values, maintenance of certified values, instructions for safety, storage, and use

Other Notes

Certified for the analytes listed below. See certificate for values and more details
Ash, Calories, Carbohydrates, Total Dietary Fiber, Fat (as the sum of fatty acids as triglycerides), Phosphorus (P), Protein
Amino acids: Alanine, Arginine, Aspartic acid, Cystine, Glutamic acid, Glycine, Histidine, Isoleucine, Leucine, Lysine, Methionine, Phenylalanine, Proline, Serine, Threonine, Tryptophan, Tyrosine, Valine
Phytochemicals: Daidzein, Daidzin, Genistein, Genistin, Glycitin
Vitamins: Pantothenic acid (Vitamin B5), Riboflavin (Vitamin B2)
Trace Elements: Calcium (Ca), Copper (Cu), Iron (Fe), Magnesium (Mg), Manganese (Mn), Potassium (K), Sodium (Na), Zinc (Zn)

Matrix Group: Nuts, Soy, Edible Oils and Fats

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service