Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

NIST3256

Green tea-containing solid oral dosage form

NIST® SRM® 3256

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

packaging

pkg of 5 x 2.5 g

manufacturer/tradename

NIST®

format

matrix material

General description

One unit of SRM 3256 contains five packets of powdered material, each containing approximately 2.5 g.

SRM 3256_cert

SRM 3256_SDS

Application

The SRM is primarily intended to validate analytical methods used to determine catechins, xanthines, andtoxic elements present in solid oral dosage forms in cluding green tea and similar matrices. Additionally, it is also suitable for ensuring quality while assigning values to in-house control materials.

Features and Benefits

  • The certified value of the SRM was determined by calculating the mass fraction values, which were obtained through two methods:
  1. Ultraviolet absorbance detection (UV)
  2. Mass spectrometry (MS)
  • Certified and reference mass fraction values for catechin,gallocatechin, gallic acid, theanine, and other elements are provided on a dry-mass basis and values are metrologically traceable to the SI unit of mass expressed as micrograms per gram.
  • The NIST certificate is provided with expiration certificate, storage, handling, use,and maintenance instructions.

Other Notes

Certified for the analytes listed below. See certificate for values and more details
Phytochemicals: (–)-Epicatechin, (–)-Epicatechin gallate, (–)-Epigallocatechin, (–)-Epigallocatechin gallate, (–)-Gallocatechin, (–)-Gallocatechin gallate, (–)-Gallocatechin gallate, (+)-Catechin, caffeine, Gallic acid, L-Theanine, Theobromine, Theophylline
Trace Metals: Arsenic (As), Cadmium (Cd), Lead (Pb), Mercury (Hg)

Matrix Group: Botanicals

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service