Skip to Content
MilliporeSigma
All Photos(1)

Documents

BCR668

Mussel tissue (trace elements)

BCR®, certified reference material

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Agency

BCR®

manufacturer/tradename

JRC

matrix

mussel

application(s)

food and beverages

format

matrix material

General description

Mussel tissue (trace elements) material consists of mussel tissue in a glass bottle containing approximately 10 g of dry powder. Information on the preparation and the certification of the rare earth elements Ce, Dy, Er, Eu, Gd, La, Lu, Nd, Pr, Sm, Tb, Tm and Y, and of the elements Th and U is given in the certification report.

Application

Mussel tissue (trace elements) has been used as a certified reference standard for the quantification of trace metals in fish samples, seafood products and mangrove sites using analytical techniques.

Analysis Note

For more information please see:
BCR668

Other Notes

Certified for the analytes listed below. See certificate for values and more details
Trace Elements: Arsenic (As), Cadmium (Cd), Cerium (Ce), Cesium (Cs), Chromium (Cr), Cobalt (Co), Dysprosium (Dy), Erbium (Er), Europium (Eu), Gadolinium (Gd), Holmium (Ho), Iron (Fe), Lanthanum (La), Lutetium (Lu), Molybdenum (Mo), Neodynium (Nd), Praseodymium (Pr), Samarium (Sm), Scandium (Sc), Terbium (Tb), Thorium (Th), Thulium, Uranium (U), Ytterbium (Yb), Yttrium (Y), Zinc (Zn)

Matrix Group: Fish and Seafood

Legal Information

BCR is a registered trademark of European Commission

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Human exposure in Italy to lead, cadmium and mercury through fish and seafood product consumption from Eastern Central Atlantic Fishing Area
Zaza S, et al.
J. Food Compos. Anal., 40, 148-153 (2015)
Multi-tool assessment of trace metals in mangroves combining sediment and clam sampling, DGT passive samplers and caged mussels
Estrada SE, et al.
The Science of the Total Environment, 574, 847-857 (2017)
Ioanna Kalantzi et al.
Journal of environmental sciences (China), 56, 300-312 (2017-06-03)
Arsenic speciation analysis was conducted on fish samples (sardine and anchovy) collected from six areas along the Greek coastline, i.e. Artemisium Straits, Thermaikos Gulf, Amvrakikos Gulf, Strymonian Gulf, Thracian Sea, and Elefsina Gulf. Total arsenic levels ranging from 11.8 to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service