Skip to Content
MilliporeSigma
All Photos(1)

Documents

NIST3233

Fortified breakfast cereal

NIST® SRM® 3233

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

packaging

pkg of 60 g ea

manufacturer/tradename

NIST®

application(s)

food and beverages

format

matrix material

General description

This Standard Reference Material (SRM) is intended primarily for validation of methods for determining proximates, sugars, dietary fiber, vitamins, elements, and amino acids in fortified breakfast cereals and similar materials. This SRM can also be used for quality assurance when assigning values to in-house reference materials. The SRM is a wheat-based fortified breakfast cereal prepared by a commercial manufacturer. A unit of SRM 3233 consists of one bottle containing approximately 60 g of material and sealed inside an aluminized pouch. For more information, please refer to the SDS and COA.

SRM 3233_cert

SRM 3233_SDS

Application


  • Vitamin D fortification in breakfast cereals: A systematic review and meta-analysis focused on the effectiveness of using fortified breakfast cereals as vehicles for improving vitamin D status, providing essential insights for nutritional science and public health strategies (Cashman and O′Neill, 2024).

  • Protein fortification for the aging population: Research discusses the design implications for protein-fortified foods, including cereals, aimed at increasing protein intake among the elderly, highlighting its significance in maintaining muscle mass and overall health (Norton et al., 2022).

  • Strategies for achieving recommended vitamin D intakes: An article outlines various fortification strategies, including the use of fortified cereals, to meet the recommended dietary intakes of vitamin D, essential for bone health and disease prevention (López-Sobaler et al., 2022).

  • Critical appraisal of ultraprocessed fortification alternatives: A critical review evaluates ultraprocessed products, including fortified cereals, as alternatives for food fortification in Latin America, discussing their impact on nutrition and public health (Kroker-Lobos et al., 2022).

  • Impact of fortified foods on iron status: A study investigates the association between the consumption of iron-fortified foods, such as cereals, and serum ferritin levels among women of childbearing age, emphasizing the importance of fortified foods in preventing iron deficiency (Nyakundi et al., 2022).

Other Notes

Certified for the analytes listed below.
Barium (Ba), Cadmium (Cd), Calcium (Ca), Copper (Cu), Iron (Fe), Magnesium (Mg), Manganese (Mn), Phosphorus (P), Potassium (K), Sodium (Na), Strontium (Sr), Zinc (Zn)
Vitamins : Thiamine (Vitamin B1), Niacinamide (Vitamin B3), Total Vitamin B3 as Niacinamide, Pantothenic Acid (Vitamin B5), Folic Acid, Total α-Tocopherol (Vitamin E)

Reference/Informational value is provided for the following analytes.
Cobalt (Co), Molybdenum (Mo), Vanadium (V), Niacin
Proximates, sugar, calories : Ash, Protein, Fat, Carbohydrates, Total Sugars, Fructose, Glucose, Maltose, Sucrose, Energy, Calories, IDF + HMW SDF, IDF, LMW SDF, HMW SDF, TDF, Alanine, Arginine, Aspartic Acid, Cystine, Glutamic Acid, Glycine, Histidine, Isoleucine, Leucine, Lysine, Methionine, Phenylalanine, Serine, Threonine, Tryptophan, Tyrosine, Valine, Aluminum (Al), Chlorine (Cl), Iodine (I), Arsenic (As)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service