Skip to Content
MilliporeSigma

58903C

Soy Hydrolysate UF Solution 50X

liquid, sterile-filtered

Synonym(s):

Soy protein enzymatic hydrolysate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.75
UNSPSC Code:
12352207
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


description

For research or for further manufacturing use

Quality Segment

sterility

sterile-filtered

form

liquid

technique(s)

cell culture | embryo: suitable

shipped in

ambient

storage temp.

2-8°C

Application

Soy Hydrolysate UF Solution 50X has been used to study the influence of the hydrolysate on the growth performance of serum-free adapted channel catfish ovary (CCO) cells.

Preparation Note

Prepared with 250 g/L soy hydrolysate ultrafiltate in tissue culture grade water.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library