Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

NIST3282

Low-Calorie Cranberry Juice Cocktail

NIST®SRM®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

form

liquid

packaging

pkg of 6 g (5 x 1.2 mL)

manufacturer/tradename

NIST®

application(s)

food and beverages

storage temp.

2-8°C

General description

This Standard Reference Material (SRM) is intended primarily for use in validating analytical methods for the determination of organic acids and nutrient elements in cranberry juice cocktails and similar matrices. This SRM can also be used for quality assurance when assigning values to in-house control materials. A unit of SRM 3282 consists of five ampoules, each containing approximately 1.2 mL of material.

SRM 3282_cert

SRM 3282_SDS

Other Notes

Certified for the analytes listed below.
Citric Acid, Malic Acid, Quinic Acid, Calcium (Ca), Copper (Cu), Magnesium (Mg), Manganese (Mn), Potassium (K), Sodium (Na)

Reference/Informational value is provided for the following analytes.
Galacturonic Acid, Glycolic Acid, Isocitric Acid, Oxalic Acid, Phosphate, Shikimic Acid, Sulfate, Iron (Fe), Zinc (Z), Fructose, Glucose, Total Sugars

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

related product

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service