Skip to Content
MilliporeSigma

03609

Ethylenediaminetetraacetic acid

BioUltra, ≥99.0% (KT)

Synonym(s):

(Ethylenedinitrilo)tetraacetic acid, EDTA, Edathamil, Ethylenedinitrilotetraacetic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
(HO2CCH2)2NCH2CH2N(CH2CO2H)2
CAS Number:
Molecular Weight:
292.24
NACRES:
NA.25
PubChem Substance ID:
eCl@ss:
39030907
UNSPSC Code:
12352116
EC Number:
200-449-4
MDL number:
Beilstein/REAXYS Number:
1716295
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


product line

BioUltra

Quality Segment

assay

≥99.0% (KT)

form

powder

reaction suitability

reagent type: chelator

impurities

insoluble matter, passes filter test

pH

2.5 (23 °C, 10 g/L)

mp

250 °C (dec.) (lit.)

solubility

0.1 M NaOH: 0.1 M at 20 °C, clear, colorless

anion traces

chloride (Cl-): ≤50 mg/kg, sulfate (SO42-): ≤100 mg/kg

cation traces

Al: ≤10 mg/kg, As: ≤0.1 mg/kg, Ba: ≤5 mg/kg, Bi: ≤5 mg/kg, Ca: ≤10 mg/kg, Cd: ≤5 mg/kg, Co: ≤5 mg/kg, Cr: ≤5 mg/kg, Cu: ≤5 mg/kg, Fe: ≤5 mg/kg, K: ≤50 mg/kg, Li: ≤5 mg/kg, Mg: ≤5 mg/kg, Mn: ≤5 mg/kg, Mo: ≤5 mg/kg, Na: ≤500 mg/kg, Ni: ≤5 mg/kg, Pb: ≤5 mg/kg, Sr: ≤5 mg/kg, Zn: ≤5 mg/kg

λ

0.1 M in 1 M NaOH

UV absorption

λ: 260 nm Amax: ≤1.00, λ: 280 nm Amax: ≤0.05

SMILES string

OC(=O)CN(CCN(CC(O)=O)CC(O)=O)CC(O)=O

InChI

1S/C10H16N2O8/c13-7(14)3-11(4-8(15)16)1-2-12(5-9(17)18)6-10(19)20/h1-6H2,(H,13,14)(H,15,16)(H,17,18)(H,19,20)

InChI key

KCXVZYZYPLLWCC-UHFFFAOYSA-N

Application

Ethylenediaminetetraacetic acid (EDTA) is a hexadentate ligand and chelating agent with the ability to sequester divalent cations such as calcium and cesium. EDTA is used in a wide range of applications where the sequestration of divalent cations is required or useful. EDTA is used in the study of artificial restriction DNA cutters.

Other Notes

Determination of anions and metal cations by single-column chromatography with EDTA as eluent using conductivity and UV detection ; Buffer component in DNA digest analysis with capillary electrophoresis


Still not finding the right product?

Explore all of our products under Ethylenediaminetetraacetic acid


signalword

Warning

Storage Class

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves

target_organs

Respiratory Tract

pictograms

Health hazardExclamation mark

Hazard Classifications

Acute Tox. 4 Inhalation - Eye Irrit. 2 - STOT RE 2 Inhalation

wgk

WGK 3



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library