Skip to Content
MilliporeSigma

Skip To

A8846

ProteoMass Angiotensin II MALDI-MS Standard

vial of 10 nmol

Synonym(s):

Angiotensin II, mass spec standard

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

NACRES:
NA.24
PubChem Substance ID:
UNSPSC Code:
41116107
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


mol wt

monoisotopic mol wt 1,045.5423 Da

Quality Level

packaging

vial of 10 nmol

SMILES string

CC[C@H](C)[C@H](NC(=O)[C@H](Cc1ccc(O)cc1)NC(=O)[C@@H](NC(=O)[C@H](CCCNC(N)=N)NC(=O)[C@@H](N)CC(O)=O)C(C)C)C(=O)N[C@@H](Cc2c[nH]cn2)C(=O)N3CCC[C@H]3C(=O)N[C@@H](Cc4ccccc4)C(O)=O

InChI

1S/C50H71N13O12/c1-5-28(4)41(47(72)59-36(23-31-25-54-26-56-31)48(73)63-20-10-14-38(63)45(70)60-37(49(74)75)22-29-11-7-6-8-12-29)62-44(69)35(21-30-15-17-32(64)18-16-30)58-46(71)40(27(2)3)61-43(68)34(13-9-19-55-50(52)53)57-42(67)33(51)24-39(65)66/h6-8,11-12,15-18,25-28,33-38,40-41,64H,5,9-10,13-14,19-24,51H2,1-4H3,(H,54,56)(H,57,67)(H,58,71)(H,59,72)(H,60,70)(H,61,68)(H,62,69)(H,65,66)(H,74,75)(H4,52,53,55)/t28-,33-,34-,35-,36-,37-,38-,40-,41-/m0/s1

InChI key

CZGUSIXMZVURDU-JZXHSEFVSA-N

Gene Information

human ... AGTR1(185)

General description

Angiotensin II (Ang II) is part of the renin-angiotensin cascade. It is an octapeptide. Ang II is present in high concentration in cardiac tissue.

Application

ProteoMass Angiotensin II MALDI-MS Standard has been used:
  • as peptide calibration standard for MALDI-MS instruments
  • as a reference standard in Liquid chromatography–mass spectrometry (LC-MS/MS) analysis for characterizing RNase and NC1 domain of collagen IV
  • as a standard in testing infrared (IR) laser desorption/ionization (IR-LDI) method

Biochem/physiol Actions

Angiotensin II (Ang II) maintains cardiac functionality by coordinating with the angiotensin II receptors. Ang II is recommended for patients with vasodilatory shock.

Legal Information

ProteoMass is a trademark of Sigma-Aldrich Co. LLC

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
A8971A8471B4181
Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

packaging

vial of 10 nmol

packaging

vial of 10 nmol

packaging

vial of 10 nmol

packaging

vial of 10 nmol

mol wt

monoisotopic mol wt 1,045.5423 Da

mol wt

(M+H+) 16,952.27 Da by calculation

mol wt

average mol wt 66,429.09 Da by calculation

mol wt

monoisotopic mol wt 756.3997 Da

Gene Information

human ... AGTR1(185)

Gene Information

-

Gene Information

-

Gene Information

-


Storage Class

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Angiotensin II and the heart: on the intracrine renin-angiotensin system
De Mello WC and Danser AH
Hypertension, 35(6), 1183-1188 (2000)
Continuous flow reduced-pressure infrared laser desorption/ionization mass spectrometry
Iguchi Y, et al.
Rapid Communications in Mass Spectrometry, 31(21), 1845-1850 (2017)
Pyridoxamine protects proteins from damage by hypohalous acids in vitro and in vivo
Madu H, et al.
Free Radical Biology & Medicine, 89(5), 83-90 (2015)



Global Trade Item Number

SKUGTIN
A8846-5X1VL04061833293232
A8846-1VL04061823921640

Questions

Reviews

No rating value

Active Filters