Skip to Content
MilliporeSigma
All Photos(1)

Documents

A9096

Sigma-Aldrich

ProteoMass Aldolase MALDI-MS Standard

vial of 10 nmol, average mol wt 39,211.28 Da by calculation

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

mol wt

average mol wt 39,211.28 Da by calculation

Quality Level

packaging

vial of 10 nmol

Application

Protein calibration standard for MALDI-MS instruments.

Legal Information

ProteoMass is a trademark of Sigma-Aldrich Co. LLC

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service