Skip to Content
MilliporeSigma

1.37028

SAFC

Glycerol

greener alternative

anhydrous (vegetable), EMPROVE® EXPERT, Ph. Eur., BP, ChP, JP, USP, ACS

Pharma Manufacturing

Synonym(s):

1,2,3-Propanetriol, Trihydroxylpropane, Protol, Glycerin, 1,2,3-Propanetriol, Glycerin

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOCH2CH(OH)CH2OH
CAS Number:
Molecular Weight:
92.09
Beilstein:
635685
MDL number:
UNSPSC Code:
12352104
EC Index Number:
200-289-5
Pricing and availability is not currently available.

grade

ACS reagent

Quality Level

Agency

BP
ChP
JP
Ph. Eur.
USP

vapor density

3.1 (vs air)

vapor pressure

<1 mmHg ( 20 °C)

product line

EMPROVE® EXPERT

Assay

≤98.0-101.0% (calculated on anhydrous substance, alkalimetric)
≥99.5% (calculated on anhydrous substance, GC)

form

liquid

autoignition temp.

698 °F

expl. lim.

2.6-11.3 % (v/v)

greener alternative product characteristics

Safer Solvents and Auxiliaries
Use of Renewable Feedstocks
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

technique(s)

electrophoresis: suitable

refractive index

n20/D 1.474 (lit.)

pH

5 (20 °C, 100 g/L in H2O)

bp

182 °C/20 mmHg (lit.)

mp

20 °C (lit.)

transition temp

flash point 199 °C

density

1.25 g/mL (lit.)

application(s)

liquid formulation
ophthalmics
parenterals
pharma/biopharma processes
pharmaceutical

greener alternative category

storage temp.

2-30°C

SMILES string

OCC(O)CO

InChI

1S/C3H8O3/c4-1-3(6)2-5/h3-6H,1-2H2

InChI key

PEDCQBHIVMGVHV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Finding the right excipient that matches your needs as well as regulatory demands can be a complicated challenge in formulation. With our application know-how and regulatory expertise, we support you in every step of development, scale-up, and production.
As part of our Emprove® Program, our raw materials are offered with extensive documentation facilitating compliance of your pharma and biopharma product, full supply chain transparency and risk mitigation. Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Green Chemistry. This product is a bio-based glycerol and is aligned with the 5th principle of Green Chemistry "Safer Solvents and Auxiliaries" and 7th principle "Use of Renewable Feedstock". Click here for more details.

Application

Glycerol can be used in a wide variety of solid, semi-solid and liquid formulations, including ophthalmic, oral solid and oral liquid dosages, tablet coatings, creams and gels. It is applied for its humectant, emollient, solvent/cosolvent, viscosity increasing, and plasticizer properties.
Glycerol anhydrous (vegetable) Emprove® Expert is low in endotoxins and bioburden, making it the right choice for your high-risk applications.

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

390.2 °F - Pensky-Martens closed cup

Flash Point(C)

199 °C - Pensky-Martens closed cup


What is the Emprove® Program?

The Emprove® Program is a system providing comprehensive and thorough documentation of our filters and single-use components, pharma raw materials, and starting materials. Four document types are included with an Emprove® Program subscription:

Material Qualification Dossier

Free of charge - Sign in to download

Material Qualification Dossier

Free of charge - Sign in to download

Quality Management Dossier (Open Access)

Free of charge - Sign in to download

Quality Management Dossier (Open Access)

Free of charge - Sign in to download

Operational Excellence Dossier

Available by subscription or for a fee

Operational Excellence Dossier

Available by subscription or for a fee

Learn more about the benefits an Emprove® Program subscription can provide
Why are these documents unavailable?
Emprove® Dossiers contain information intended to support the registration process and risk assessment activities of pharmaceutical producers and contain confidential information. Please contact us if you would like access.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Articles

This article outlines critical factors in long-term stability of proteins, and how different stabilizers protect again mechanical and thermal stress.

This article outlines critical factors in long-term stability of proteins, and how different stabilizers protect again mechanical and thermal stress.

This article outlines critical factors in long-term stability of proteins, and how different stabilizers protect again mechanical and thermal stress.

This article outlines critical factors in long-term stability of proteins, and how different stabilizers protect again mechanical and thermal stress.

See All

Related Content

Developing formulations specifically for infants and children is increasingly important. To help you master these challenges effectively, we offer an extensive portfolio of well-established high-quality excipients and APIs for pediatric pharmaceutical formulations that are proven in practice.

Developing formulations specifically for infants and children is increasingly important. To help you master these challenges effectively, we offer an extensive portfolio of well-established high-quality excipients and APIs for pediatric pharmaceutical formulations that are proven in practice.

Developing formulations specifically for infants and children is increasingly important. To help you master these challenges effectively, we offer an extensive portfolio of well-established high-quality excipients and APIs for pediatric pharmaceutical formulations that are proven in practice.

Developing formulations specifically for infants and children is increasingly important. To help you master these challenges effectively, we offer an extensive portfolio of well-established high-quality excipients and APIs for pediatric pharmaceutical formulations that are proven in practice.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service