Skip to Content
MilliporeSigma

8.18709

Glycerol

greener alternative

anhydrous for synthesis

Synonym(s):

1,2,3-Propanetriol, Trihydroxylpropane, Protol, Glycerin, 1,2,3-Propanetriol, Glycerin

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
HOCH2CH(OH)CH2OH
CAS Number:
Molecular Weight:
92.09
UNSPSC Code:
12352104
NACRES:
NA.22
EC Index Number:
200-289-5
MDL number:
Beilstein/REAXYS Number:
635685
Assay:
≥99% (calc. on anhydr. substance, GC)
Grade:
synthesis grade
Technique(s):
electrophoresis: suitable
Bp:
182 °C/20 mmHg (lit.)
Vapor pressure:
<1 mmHg ( 20 °C)
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

synthesis grade

Quality Segment

vapor density

3.1 (vs air)

vapor pressure

<1 mmHg ( 20 °C)

assay

≥99% (calc. on anhydr. substance, GC)

form

liquid

autoignition temp.

698 °F

potency

12600 mg/kg LD50, oral (Rat), >18700 mg/kg LD50, skin (Rabbit)

expl. lim.

2.6-11.3 % (v/v)

greener alternative product characteristics

Safer Solvents and Auxiliaries
Design for Degradation
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

technique(s)

electrophoresis: suitable

refractive index

n20/D 1.474 (lit.)

pH

5 (20 °C, 100 g/L in H2O)

bp

182 °C/20 mmHg (lit.)

mp

20 °C (lit.)

transition temp

flash point 199 °C

density

1.25 g/mL (lit.)

greener alternative category

storage temp.

2-30°C

SMILES string

OCC(O)CO

InChI

1S/C3H8O3/c4-1-3(6)2-5/h3-6H,1-2H2

InChI key

PEDCQBHIVMGVHV-UHFFFAOYSA-N

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of the 12 Principles of Green Chemistry. Glycerol, with its high boiling point, low flammability, and polarity, is a standout choice. As a non-toxic and biodegradable compound, it aligns perfectly with the principles of ′Safer Solvents and Auxiliaries′ and ′Design for Degradation′. Click here for more information.

Analysis Note

Assay (GC, calc. on anhydr. substance): ≥ 99 %
Identity (IR-spectrum): passes test
Density (d 20/4): 1.259 - 1.263 g/cm³
Water: ≤ 0.5 %


Still not finding the right product?

Explore all of our products under Glycerol


Storage Class

10 - Combustible liquids

wgk

WGK 1

flash_point_f

390.2 °F - Pensky-Martens closed cup

flash_point_c

199 °C - Pensky-Martens closed cup



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library