Skip to Content
MilliporeSigma
Glycerol BioRenewable, ACS reagent, ≥99.5%

Glycerol

Glycerol BioRenewable, ACS reagent, ≥99.5%
Synonym(s):
1,2,3-Propanetriol, Glycerin, Glycerol
Linear Formula:
HOCH2CH(OH)CH2OH
CAS Number:
Molecular Weight:
92.09

Products

Product Category
Brand
Sustainability
Boiling Point (°C)
Feature
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Research Area
Quality Segment
Application
Special Grade
Technique
Showing 1-20 of 29
Page 1 of 2
Page 1 of 2