Skip to Content
MilliporeSigma

1.00013

Acetone

EMPROVE® ESSENTIAL, Ph. Eur., BP, JPE, NF

Synonym(s):

Dimethyl ketone, Propanone, 2-Propanone

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
CH3COCH3
CAS Number:
Molecular Weight:
58.08
EC Index Number:
200-662-2
UNSPSC Code:
12352115
Beilstein/REAXYS Number:
635680
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

agency

BP, JPE, NF, Ph. Eur.

vapor density

2 (vs air)

vapor pressure

184 mmHg ( 20 °C)

description

EMPROVE® ESSENTIAL Ph Eur,BP,JPE,NF

product line

EMPROVE® ESSENTIAL

assay

≥99% (GC)

form

liquid

autoignition temp.

465 °C (DIN 51794)

expl. lim.

13.2 %

technique(s)

API processing | polymorph screening: suitable

refractive index

n20/D 1.359 (lit.)

pH

5-6 (20 °C, 395 g/L in H2O)

capacity

1000  g/kg water absorbing capacity

bp

56 °C/760 mmHg (lit.)

mp

−94 °C (lit.)

transition temp

flash point <-20 °C

density

0.791 g/mL at 25 °C (lit.)

application(s)

liquid formulation
pharma/biopharma processes
pharmaceutical

format

neat

storage temp.

2-30°C

SMILES string

CC(C)=O

InChI

1S/C3H6O/c1-3(2)4/h1-2H3

InChI key

CSCPPACGZOOCGX-UHFFFAOYSA-N

General description

Finding the right excipient that matches your needs as well as regulatory demands can be a complicated challenge in formulation. With our application know-how and regulatory expertise, we support you in every step of development, scale-up, and production.
As part of our Emprove® Program, our raw materials are offered with extensive documentation facilitating compliance of your pharma and biopharma product, full supply chain transparency and risk mitigation. Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.

Application

Acetone is used as solvent in a variety of pharmaceutical production processes, e. g. tablet coating, granulation, and microsphere production. It is also applied as solvent or co-solvent in topical formulations.
Acetone′s luminesence intensity is dependent upon the solution components . The absorption of UV light by acetone, results in its photolysis and the production of radials .

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?

Explore all of our products under Acetone


pictograms

FlameExclamation mark

signalword

Danger

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

target_organs

Central nervous system

supp_hazards

Storage Class

3 - Flammable liquids

wgk

WGK 1

flash_point_f

1.4 °F - closed cup

flash_point_c

-17.0 °C - closed cup



Stay compliant with GMP using the Emprove® Program

Accelerate qualification, maintain compliance, and save time with Emprove® Dossiers.

Download Dossiers

Note: you will be brought to EmproveSuite.com. Your username and password are separate from SigmaAldrich.com and may differ.

Register for free and access Emprove® Dossiers within minutes. Upgrade your subscription plan to unlock advanced and customized content in the Emprove® Suite

Not finding what you are looking for? Visit the Emprove® Program Help page.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library