Skip to Content
MilliporeSigma
All Photos(1)

Documents

G2289

Sigma-Aldrich

Glycerol

meets USP testing specifications

Synonym(s):

Glycerol, 1,2,3-Propanetriol, Glycerin

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOCH2CH(OH)CH2OH
CAS Number:
Molecular Weight:
92.09
Beilstein:
635685
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.21

product name

Glycerin, meets USP testing specifications

biological source

plant (palm)

Quality Level

Agency

USP/NF
meets USP testing specifications

vapor density

3.1 (vs air)

vapor pressure

<1 mmHg ( 20 °C)

Assay

99.0-101.0% anhydrous basis

form

viscous liquid

autoignition temp.

698 °F

technique(s)

cryopreservation: suitable
immunofluorescence: suitable

impurities

<0.01% Ash
<5% water (Karl Fischer)

refractive index

n20/D 1.474 (lit.)

bp

182 °C/20 mmHg (lit.)

mp

20 °C (lit.)

solubility

water: miscible

density

1.25 g/mL (lit.)

application(s)

agriculture
life science and biopharma
pharmaceutical (small molecule)
sample preparation

SMILES string

OCC(O)CO

InChI

1S/C3H8O3/c4-1-3(6)2-5/h3-6H,1-2H2

InChI key

PEDCQBHIVMGVHV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Glycerol, also referred to as glycerin or 1,2,3-propanetriol, is a remarkably versatile compound with its three hydroxyl groups, enabling strong solubility in water and rendering it hygroscopic. This simple but widely accessible substance has multifaceted applications, serving as both a carbon source and a precursor in diverse chemical and biological processes. In the realm of biotechnology, glycerol is an indispensable resource for various microorganisms, allowing them to produce a wide array of bioproducts when used as the sole carbon and energy source. Furthermore, glycerol finds extensive utility in sample preparation, where it enhances sample density and guards against freezing.Glycerol′s significance transcends biotechnology as it excels in sample preparation, such as electrophoresis gel creation. Concentrations of 5% to 10% glycerol are deployed to heighten sample density, ensuring precise layering at the base of the gel. This versatile compound also aids in casting gradient gels and acts as a stabilizer for proteins while playing a pivotal role in storage buffers. It is also a favored choice for enzyme concentration and storage, effectively preventing protein back-diffusion into the buffer. In the context of cell culture, glycerol proves its worth as a cryoprotectant, lowering the freezing point of the culture medium and shielding cells from ice crystal damage during -80°C freezing and storage.Glycerol plays a crucial role in sample preservation and storage in the field of biochemical research. Freezing bacterial cultures alone can have detrimental effects, resulting in dehydration and protein denaturation due to ice crystal formation. By incorporating glycerol, these issues are mitigated, ensuring the effective preservation of bacterial cultures. Storing bacterial cultures in glycerol stocks ensures stability, preventing harm to cell membranes and maintaining cell viability, thus enabling long-term storage. In the context of laboratory research, the preservation of sample integrity is paramount, and glycerol′s role in this process is instrumental, especially when stored in Ultra-low Temperature Freezers at -80°C to maintain sample integrity and ensure their suitability for extensive analysis and research.

Application

Glycerol has been used

  • as a cryoprotectant in the sample medium for research related to plant physiology
  • as an optical clearing agent in a study about harmonic generation microscopy

Features and Benefits

  • Can be used in Metabolomics and Biochemical research
  • High-quality compound suitable for multiple research applications

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

390.2 °F - Pensky-Martens closed cup

Flash Point(C)

199 °C - Pensky-Martens closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Starch-Based Superabsorbent Enhances the Growth and Physiological Traits of Ornamental Shrubs
Pacholczak A et al.
Agriculture Science Developments, 13, 1893-1893 (2023)
Jia-Hong Lai et al.
Scientific reports, 11(1), 329-329 (2021-01-13)
Imaging depth and quality of optical microscopy can be enhanced by optical clearing. Here we investigate the optical clearing of the ex vivo human skin by 50% glycerol topical application, which is allowed for cosmetic usage. Harmonic generation microscopy, by
Ruifang Wang et al.
Autophagy, 15(7), 1163-1181 (2019-02-12)
Influenza A virus can evade host innate immune response that is involved in several viral proteins with complicated mechanisms. To date, how influenza A M2 protein modulates the host innate immunity remains unclear. Herein, we showed that M2 protein colocalized
Shelley L Forrest et al.
The Journal of comparative neurology, 522(17), 3900-3927 (2014-07-22)
Artemin is a member of the glial cell line-derived neurotrophic factor (GDNF) family that has been strongly implicated in development and regeneration of autonomic nerves and modulation of nociception. Whereas other members of this family (GDNF and neurturin) primarily target
Nazish Ahmed et al.
Tissue engineering. Part A, 20(15-16), 2224-2233 (2014-03-13)
One of the factors preventing clinical application of regenerative medicine to degenerative cartilage diseases is a suitable source of cells. Chondrocytes, the only cell type of cartilage, grown in vitro under culture conditions to expand cell numbers lose their phenotype

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service