Skip to Content
MilliporeSigma

Skip To

1.00995

2-Propanol

EMPROVE® ESSENTIAL, Ph. Eur., BP, ChP, JP, USP

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

EC Index Number:
200-661-7
UNSPSC Code:
12191502
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

agency

BP, ChP, JP, Ph. Eur., USP

vapor pressure

43 hPa ( 20 °C)

product line

EMPROVE® ESSENTIAL

assay

≥99.5% (GC)

form

liquid

technique(s)

API processing | polymorph screening: suitable

pH

7 (20 °C)

transition temp

flash point 12 °C

density

0.786 g/cm3 at 20 °C

application(s)

liquid formulation
pharma/biopharma processes
pharmaceutical
semi-solid formulation

storage temp.

2-25°C

General description

Finding the right excipient that matches your needs as well as regulatory demands can be a complicated challenge in formulation. With our application know-how and regulatory expertise, we support you in every step of development, scale-up, and production.
As part of our Emprove® Program, our raw materials are offered with extensive documentation facilitating compliance of your pharma and biopharma product, full supply chain transparency and risk mitigation. Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.

Application

2-Propanol is used as a solvent in a variety of pharmaceutical production processes, e. g. tablet coating, granulation, and microsphere production. It is also applied as a solvent or co-solvent in topical formulations.

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany


Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Codes

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Storage Class

3 - Flammable liquids

Flash Point (°F)

53.6 °F - closed cup

Flash Point (°C)

12.0 °C - closed cup

Target Organs

Central nervous system



Stay compliant with GMP using the Emprove® Program

Accelerate qualification, maintain compliance, and save time with Emprove® Dossiers.

Download Dossiers

Note: you will be brought to EmproveSuite.com. Your username and password are separate from SigmaAldrich.com and may differ.

Register for free and access Emprove® Dossiers within minutes. Upgrade your subscription plan to unlock advanced and customized content in the Emprove® Suite

Not finding what you are looking for? Visit the Emprove® Program Help page.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Difficulty Finding Your Product Or Lot/Batch Number?

Product numbers are combined with Pack Sizes/Quantity when displayed on the website (example: T1503-25G). Please make sure you enter ONLY the product number in the Product Number field (example: T1503).

Example:

T1503
Product Number
-
25G
Pack Size/Quantity

Additional examples:

705578-5MG-PW

PL860-CGA/SHF-1EA

MMYOMAG-74K-13

1000309185

enter as 1.000309185)

Having trouble? Feel free to contact Technical Service for assistance.



Questions

Reviews

No rating value

Active Filters