Skip to Content
Merck

Skip To

MSCAL3

ProteoMass Protein MALDI-MS Calibration Kit

Protein MALDI MS Calibration Standard

Synonym(s):

Protein Mass Spectrometry Calibrators

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

NACRES:
NA.24
UNSPSC Code:
12161503
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


product line

ProteoMass

Quality Level

quality

Protein MALDI MS Calibration Standard

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

storage temp.

15-25°C

Application

This kit features a convenient selection of pre-qualified standard proteins, matrices, and solvents for calibrating and testing matrix assisted laser desorption ionization (MALDI) mass spectrometers. Configured specifically for applications in the mass range of 5 to 66 kDa, MSCAL3 is ideal for most protein studies. Applications include calibration, tuning, and sensitivity testing of MALDI instruments from all manufacturers.
ProteoMass Protein MALDI-MS Calibration Kit has been used as a mass calibration standard for MALDI-TOF MS (matrix assisted laser desorption ionization-time of flight mass spectrometry) analysis.[1]

Legal Information

ProteoMass is a trademark of Sigma-Aldrich Co. LLC

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
MSCAL1MSCAL2C8857
product line

ProteoMass

product line

ProteoMass

product line

ProteoMass

product line

ProteoMass

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

quality

Protein MALDI MS Calibration Standard

quality

Peptide and Protein MALDI MS Calibration Standard

quality

Peptide MALDI MS Calibration Standard

quality

-

storage temp.

15-25°C

storage temp.

-

storage temp.

15-25°C

storage temp.

-


Kit Components Only

Product No.
Description

  • Sinapinic acid, Use tested for MALDI-TOF Mass Spec. 8 x 10

Kit Components Also Available Separately

Product No.
Description
SDS & Pricing

  • ProteoMass Insulin MALDI-MS Standard, vial of 10 nmol, monoisotopic mol wt 5,729.6087 Da 2 x 10
    SDS

  • ProteoMass Cytochrome c MALDI-MS Standard, vial of 10 nmol, (M+H+) 12,361.96 Da by calculation 2 x 10
    SDS

  • ProteoMass Apomyoglobin MALDI-MS Standard, vial of 10 nmol, (M+H+) 16,952.27 Da by calculation 2 x 10
    SDS

  • ProteoMass Aldolase MALDI-MS Standard, vial of 10 nmol, average mol wt 39,211.28 Da by calculation 2 x 10
    SDS

  • ProteoMass Albumin MALDI-MS Standard, vial of 10 nmol, average mol wt 66,429.09 Da by calculation 2 x 10
    SDS

  • Acetonitrile 30 mL
    SDS

pictograms

FlameExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

3 - Flammable liquids

wgk

WGK 3



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Wipawadee Sianglum et al.
PloS one, 6(2), e16628-e16628 (2011-02-18)
The ethanolic extract from Rhodomyrtus tomentosa leaf exhibited good antibacterial activities against both methicillin-resistant Staphylococcus aureus (MRSA) and S. aureus ATCC 29213. Its minimal inhibitory concentration (MIC) values ranged from 31.25-62.5 µg/ml, and the minimal bactericidal concentration (MBC) was 250
Marta Martínez-Júlvez et al.
The FEBS journal, 279(23), 4306-4317 (2012-10-09)
The RdxA oxygen-insensitive nitroreductase of the human gastric pathogen Helicobacter pylori is responsible for the susceptibility of this organism to the redox active prodrug metronidazole [2-(2-methyl-5-nitro-1H-imidazol-1-yl)ethanol]. Loss-of-function mutations in rdxA are primarily responsible for resistance to this therapeutic. RdxA exhibits potent
Tyler A Zimmerman et al.
Analytical chemistry, 83(15), 6090-6097 (2011-06-23)
In-source decay (ISD) fragmentation as combined with matrix-assisted laser desorption/ionization (MALDI) mass spectrometry allows protein sequencing directly from mass spectra. Acquisition of MALDI-ISD mass spectra from tissue samples is achieved using an appropriate MALDI matrix, such as 1,5-diaminonaphthalene (DAN). Recent



Global Trade Item Number

SKUGTIN
MSCAL3-1KT04061834066880
MSCAL3-5X1KT04061833652664

Questions

Reviews

No rating value

Active Filters