Skip to Content
Merck
All Photos(1)

Key Documents

B4181

Sigma-Aldrich

ProteoMass Bradykinin Fragment 1-7 MALDI-MS Standard

vial of 10 nmol, monoisotopic mol wt 756.3997 Da

Synonym(s):

Bradykinin fragment 1-7, mass spec standard

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

mol wt

monoisotopic mol wt 756.3997 Da

Quality Level

packaging

vial of 10 nmol

SMILES string

NC(CCCNC(N)=N)C(=O)N1CCCC1C(=O)N2CCCC2C(=O)NCC(=O)NC(Cc3ccccc3)C(=O)NC(CO)C(=O)N4CCCC4C(O)=O

InChI

1S/C35H52N10O9/c36-22(10-4-14-39-35(37)38)31(50)44-16-6-12-26(44)33(52)43-15-5-11-25(43)30(49)40-19-28(47)41-23(18-21-8-2-1-3-9-21)29(48)42-24(20-46)32(51)45-17-7-13-27(45)34(53)54/h1-3,8-9,22-27,46H,4-7,10-20,36H2,(H,40,49)(H,41,47)(H,42,48)(H,53,54)(H4,37,38,39)

InChI key

CRROPKNGCGVIOG-UHFFFAOYSA-N

General description

ProteoMass Bradykinin Fragment 1-7 MALDI-MS Standard is required to calibrate and test matrix assisted laser desorption ionization (MALDI) mass spectrometers.
1 vial contains 10nmols.

Application

ProteoMass Bradykinin Fragment 1-7 MALDI-MS Standard has been used as an internal standard for MALDI-TOF-MS (matrix assisted laser desorption ionization-time of flight mass spectrometry) analysis. It has been used as a peptide analyte to study the influence of solvent on MALDI-TOF-MS analysis.

Legal Information

ProteoMass is a trademark of Sigma-Aldrich Co. LLC

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effect of solvent on ionization efficiency in matrix-assisted laser desorption/ionization mass spectrometry of peptides.
Hazama H, et al.
Chemical Physics, 419, 196-199 (2013)
Proteomic identification of antioxidant peptides from 400 to 2500Da generated in Spanish dry-cured ham contained in a size-exclusion chromatography fraction.
Mora L, et al.
Food Research International, 56, 68-76 (2014)

Articles

One of the most important aspects of our ultra-pure MALDI matrix substances is their ability to dissolve rapidly and completely; a brief vortex mixing is typically sufficient.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service