A8971
ProteoMass™ Apomyoglobin MALDI-MS Standard
vial of 10 nmol, (M+H+) 16,952.27 Da by calculation
Synonym(s):
Apomyoglobin, mass spec standard
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
General description
ProteoMass™ Apomyoglobin MALDI-MS Standard is required to calibrate and test matrix assisted laser desorption ionization (MALDI) mass spectrometers.
Application
ProteoMass™ Apomyoglobin MALDI-MS Standard has been used as an internal standard for MALDI-TOF-MS (matrix assisted laser desorption ionization-time of flight mass spectrometry) analysis.
Legal Information
ProteoMass is a trademark of Sigma-Aldrich Co. LLC
related product
Product No.
Description
Pricing
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Study of papain-cystatin interaction by intensity fading MALDI-TOF-MS.
Shabab M, et al.
The Protein Journal, 27, 7-12 (2008)
Chuchu Zhang et al.
Proceedings of the National Academy of Sciences of the United States of America, 114(12), E2524-E2532 (2017-03-08)
Pain-producing animal venoms contain evolutionarily honed toxins that can be exploited to study and manipulate somatosensory and nociceptive signaling pathways. From a functional screen, we have identified a secreted phospholipase A2 (sPLA2)-like protein, BomoTx, from the Brazilian lancehead pit viper
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service