Merck
  • Comparison of rheological properties, follicular penetration, drug release, and permeation behavior of a novel topical drug delivery system and a conventional cream.
Comparison of rheological properties, follicular penetration, drug release, and permeation behavior of a novel topical drug delivery system and a conventional cream.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V (2014-12-03)
Andreas Lauterbach, Christel C Müller-Goymann
ABSTRACT
A novel adapalene-loaded solid lipid microparticle (SLMA) dispersion as a topical drug delivery system (TDDS) for follicular penetration has been introduced. The objective of the present study was to investigate the rheological properties, the follicular penetration with differential tape stripping on porcine ear skin, the drug release in sebum and stratum corneum (SC) lipid mixtures, and the permeation behavior across human SC in comparison with a commercially available cream as standard. Physicochemical characterization reveals that adapalene is homogeneously distributed within the SLMA dispersion and chemically stable for at least 24 weeks. The SLMA dispersion shows a lower complex viscosity at 20 °C and a higher one at 32 °C than the cream, while the phase angle of the dispersion is always larger at both temperatures. Both formulations feature an equivalent potential for follicular penetration and deposition. However, the superiority of the SLMA dispersion is based on the preferential drug release in sebum while there is no or just a slight release in SC lipids and no permeation for both formulations. Due to the similarity of the glyceride matrix of the SLMA to sebum components, a targeted drug delivery into sebum and thereby an increased follicular penetration may be facilitated.
Product Description
Supelco
Cholesterol solution, certified reference material, 10 mg/mL in chloroform
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, aqueous solution, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Supelco
Methyl Paraben, PESTANAL®, analytical standard
Supelco
Palmitic acid, analytical standard
Supelco
Acetic acid, analytical standard
Sigma-Aldrich
Cholesterol, from lanolin, ≥99.0% (GC)
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Tetrahydrofuran, anhydrous, contains 250 ppm BHT as inhibitor, ≥99.9%
Supelco
Tetrahydrofuran, HPLC grade, ≥99.9%, inhibitor-free
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
trans,trans-2,4-Hexadienoic acid potassium salt, ReagentPlus®, 99%
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Tetrahydrofuran, anhydrous, ≥99.9%, inhibitor-free
Sigma-Aldrich
Palmitic acid, ≥98%, FCC, FG
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Sigma-Aldrich
Oleyl oleate, ≥99%
Sigma-Aldrich
Palmitic acid, ≥98% palmitic acid basis (GC)
Supelco
Tetrahydrofuran, analytical standard
Supelco
Tetrahydrofuran, Selectophore, ≥99.5%
Sigma-Aldrich
Palmitic acid, ≥99%
Sigma-Aldrich
Palmitic acid, BioXtra, ≥99%
Sigma-Aldrich
Cholesterol, tested according to Ph. Eur.
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
Supelco
Methylparaben, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Palmitic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Palmitic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Palmitic acid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Acetic acid solution, suitable for HPLC