Skip to Content
Merck
All Photos(3)

Key Documents

186562

Sigma-Aldrich

Tetrahydrofuran

≥99.9%, anhydrous, contains 250 ppm BHT as inhibitor

Synonym(s):

THF, Butylene oxide, Oxolane, Tetramethylene oxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H8O
CAS Number:
Molecular Weight:
72.11
Beilstein:
102391
EC Number:
MDL number:
UNSPSC Code:
12191501
PubChem Substance ID:
NACRES:
NA.21
grade:
anhydrous
bp:
65-67 °C (lit.)
vapor pressure:
114 mmHg ( 15 °C)
143 mmHg ( 20 °C)

Product Name

Tetrahydrofuran, anhydrous, contains 250 ppm BHT as inhibitor, ≥99.9%

grade

anhydrous

Quality Level

vapor density

2.5 (vs air)

vapor pressure

114 mmHg ( 15 °C)
143 mmHg ( 20 °C)

Assay

≥99.9%

form

liquid

autoignition temp.

610 °F

contains

250 ppm BHT as inhibitor

expl. lim.

1.8-11.8 %

impurities

≤0.005% peroxides (as H2O2)
<0.002% water (All units except 100 mL)
<0.005% water (100 mL pkg)

refractive index

n20/D 1.407 (lit.)

pH

~7

bp

65-67 °C (lit.)

mp

−108 °C (lit.)

solubility

H2O: soluble

density

0.889 g/mL at 25 °C (lit.)

SMILES string

C1CCOC1

InChI

1S/C4H8O/c1-2-4-5-3-1/h1-4H2

InChI key

WYURNTSHIVDZCO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Tetrahydrofuran (THF) is a saturated cyclic ether mainly used as an organic solvent. On long term storage it forms organic peroxides. This process can be suppressed by adding butylated hydroxytoluene (BHT) as a stabilizer. BHT removes the free radicals required for the peroxide formation. It constitutes the key fragment of various natural products (polyether antibiotics). THF forms double hydrate with hydrogen sulfide, crystal structure of this double hydrate has been investigated by three-dimensional single-crystal studies.

Application

Dry tetrahydrofuran (THF) has been used in the synthesis of dichloro{bis[1,1′,1′′-(phosphinetriyl)tripiperidine]}palladium [(P(NC5H10)3)2Pd(Cl)2], which is widely used in the Heck reaction as a catalyst. It has been employed in the reaction medium of the Heck cross-coupling reaction of various aryl bromides and olefins.

Other Notes

Greener alternatives are available for many THF applications, 2-Methyltetrahydrofuran (414247) and Cyclopentyl methyl ether (675970)

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

-6.2 °F - closed cup

Flash Point(C)

-21.2 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Mizoroki-Heck Cross-coupling Reactions Catalyzed by Dichloro {bis [1, 1', 1''-(phosphinetriyl) tripiperidine]} palladium Under Mild Reaction Conditions.
Oberholzer M and Frech CM
Journal of Visualized Experiments, 85, e51444-e51444 (2014)
Mauro Niso et al.
Chembiochem : a European journal of chemical biology, 16(7), 1078-1083 (2015-03-11)
Although sigma-2 (σ2 ) receptors are still enigmatic proteins, they are promising targets for tumor treatment and diagnosis. With the aim of clarifying their role in oncology, we developed a σ2 -selective fluorescent tracer (compound 5) as a specific tool
Francesco Berardi et al.
Journal of medicinal chemistry, 52(23), 7817-7828 (2009-10-22)
sigma(2)-Agonist 1-cyclohexyl-4-[3-(5-methoxy-1,2,3,4-tetrahydronaphthalen-1-yl)propyl]piperazine (7, PB 28), which proved to revert doxorubicin resistance in breast cancer cells, was taken as a template to prepare new analogs. One of the two basic N-atoms was alternatively replaced by a methine or converted into an
Maria Laura Pati et al.
Pharmacological research, 117, 67-74 (2016-12-23)
A controversial relationship between sigma-2 and progesterone receptor membrane component 1 (PGRMC1) proteins, both representing promising targets for the therapy and diagnosis of tumors, exists since 2011, when the sigma-2 receptor was reported to be identical to PGRMC1. Because a
Nicola Antonio Colabufo et al.
Naunyn-Schmiedeberg's archives of pharmacology, 370(2), 106-113 (2004-08-24)
To establish the activity of sigma ligands at sigma1 and sigma2 receptor, we chose two tumour cell lines, the human SK-N-SH neuroblastoma and the rat C6 glioma lines, which express sigma2 receptors at a high density and sigma1 receptors in

Articles

A description of stabilizer systems for solvents including esters, ethyl ester, tetrahydrofuran, chlorinated solvents, chloroform and dichloromethane.

A description of stabilizer systems for solvents including esters, ethyl ester, tetrahydrofuran, chlorinated solvents, chloroform and dichloromethane.

A description of stabilizer systems for solvents including esters, ethyl ester, tetrahydrofuran, chlorinated solvents, chloroform and dichloromethane.

A description of stabilizer systems for solvents including esters, ethyl ester, tetrahydrofuran, chlorinated solvents, chloroform and dichloromethane.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service