Skip to Content
Merck
All Photos(3)

Key Documents

W200603

Sigma-Aldrich

Acetic acid

greener alternative

natural, ≥99.5%, FG

Synonym(s):

Glacial acetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CO2H
CAS Number:
Molecular Weight:
60.05
FEMA Number:
2006
Beilstein:
506007
EC Number:
Council of Europe no.:
0002
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
8.002
NACRES:
NA.21
grade:
FG
Halal
Kosher
natural
food allergen:
no known allergens

grade

FG
Halal
Kosher
natural

Quality Level

Agency

meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117
FDA 21 CFR 173.370
FDA 21 CFR 184.1005

vapor density

2.07 (vs air)

vapor pressure

15.2 hPa ( 20.0 °C)
73.3 hPa ( 50.0 °C)

Assay

≥99.5%

form

liquid

autoignition temp.

800 °F

shelf life

Expiration date period - 5 years

expl. lim.

16 %, 92 °F
4 %, 59 °F

greener alternative product characteristics

Less Hazardous Chemical Syntheses
Use of Renewable Feedstocks
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

color

colorless

refractive index

n20/D 1.371 (lit.)

pH

2.5 (20 °C, 50 g/L)

bp

117-118 °C (lit.)

mp

16.2 °C (lit.)

density

1.04 g/mL at 25 °C (lit.)

cation traces

As: ≤3 ppm
Cd: ≤1 ppm
Hg: ≤1 ppm
Pb: ≤1 ppm

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

greener alternative category

Organoleptic

sour

SMILES string

[F2C(F2C)13F3C]C(O)=O

InChI

1S/C2H4O2/c1-2(3)4/h1H3,(H,3,4)

InChI key

QTBSBXVTEAMEQO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Acetic acid is an organic acid mainly used as an acidulating agent and flavoring agent. It is one of the main volatile constituents of vinegar and pyroligneous acid. In combination with leavening agents, it produces carbon dioxide from sodium bicarbonate. Acetic acid is used as a flavoring agent in ice-creams, nonalcoholic beverages and baked goods.
We are committed to bringing you Greener Alternative Products, which adhere to one of the four categories of Greener Alternatives . This product is a Biobased products, showing key improvements in Green Chemistry Principles “Less Hazardous Chemical Syntheses” and “Use of Renewable Feedstock”.

Application


  • Investigation of Potential Gut Health Biomarkers in Broiler Chicks Challenged by Campylobacter jejuni and Submitted to a Continuous Water Disinfection Program.: This study explores the effects of acetic acid-based disinfection programs on gut health biomarkers in broiler chicks, aiming to improve their resistance to Campylobacter jejuni infection (Mantzios et al., 2024).

  • Distribution Characteristics of Low-Molecular-Weight Organic Acids in Reclaimed Soil Filled with Fly Ash: A Study.: This research investigates the presence and distribution of acetic acid and other low-molecular-weight organic acids in reclaimed soils, focusing on their environmental impact and potential uses in soil remediation (Zheng et al., 2024).

  • Antibiotic Cocktail Effects on Intestinal Microbial Community, Barrier Function, and Immune Function in Early Broiler Chickens.: This research evaluates the influence of acetic acid and other antibiotics on the intestinal health and immune function of broiler chickens, emphasizing its benefits in early poultry development (Abbas et al., 2024).

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1A

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

102.2 °F - closed cup

Flash Point(C)

39 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Fenaroli G
Fenaroli's Handbook of Flavor Ingredients, 5-5 (1971)
Bender DA and Bender AE
Benders? Dictionary of Nutrition and Food Technology (1999)
Burdock GA
Encyclopedia of Food and Color Additives, 1, 13-15 (1997)
IgoeY Rs, et al
Dictionary of Food ingredients, 3-3 (1999)
Takashi Taniguchi et al.
Arthritis & rheumatology (Hoboken, N.J.), 67(2), 517-526 (2014-11-12)
Fli-1, a potential predisposing factor for systemic sclerosis (SSc), is constitutively down-regulated in the lesional skin of patients with SSc by an epigenetic mechanism. To investigate the impact of Fli-1 deficiency on the induction of an SSc phenotype in various

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service