Recommended Products
description
13C-depleted
isotopic purity
99.9 atom % 12C
Assay
99% (CP)
bp
117-118 °C (lit.)
mp
16.2 °C (lit.)
density
1.049 g/mL at 25 °C
mass shift
depleted
SMILES string
[12CH3][12C](O)=O
InChI
1S/C2H4O2/c1-2(3)4/h1H3,(H,3,4)/i1+0,2+0
InChI key
QTBSBXVTEAMEQO-ZCWHFVSRSA-N
Packaging
This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1A
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
104.0 °F - closed cup
Flash Point(C)
40 °C - closed cup
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
The Journal of clinical investigation, 125(7), 2592-2608 (2015-05-27)
The integrated stress response (ISR) is a critical mediator of cancer cell survival, and targeting the ISR inhibits tumor progression. Here, we have shown that activating transcription factor 4 (ATF4), a master transcriptional effector of the ISR, protects transformed cells
American journal of physiology. Regulatory, integrative and comparative physiology, 308(2), R123-R130 (2014-11-21)
In healthy individuals, postexercise protein supplementation increases muscle protein anabolism. In patients with muscular dystrophies, aerobic exercise improves muscle function, but the effect of exercise on muscle protein balance is unknown. Therefore, we investigated 1) muscle protein balance before, during
The Journal of neuroscience : the official journal of the Society for Neuroscience, 35(32), 11281-11291 (2015-08-14)
After ischemic stroke, various damage-associated molecules are released from the ischemic core and diffuse to the ischemic penumbra, activating microglia and promoting proinflammatory responses that may cause damage to the local tissue. Here we demonstrate using in vivo and in
Oncology reports, 34(1), 279-287 (2015-05-23)
Melatonin is a small indole produced by the pineal gland and other tissues, and has numerous functions that aid in the maintenance of the whole body homeostasis, ranging from the regulation of circadian rhythms and sleep to protection from oxidative
Molecular biology and evolution, 32(7), 1862-1879 (2015-03-17)
In most eukaryotes, centromere is determined by the presence of the centromere-specific histone variant CenH3. Two types of chromosome morphology are generally recognized with respect to centromere organization. Monocentric chromosomes possess a single CenH3-containing domain in primary constriction, whereas holocentric
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service