Skip to Content
Merck
All Photos(6)

Documents

C8667

Sigma-Aldrich

Cholesterol

Sigma Grade, ≥99%

Synonym(s):

3β-Hydroxy-5-cholestene, 5-Cholesten-3β-ol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H46O
CAS Number:
Molecular Weight:
386.65
Beilstein:
1915888
EC Number:
MDL number:
UNSPSC Code:
12352111
eCl@ss:
39023139
PubChem Substance ID:
NACRES:
NA.77

biological source

sheep wool

grade

Sigma Grade

Assay

≥99%

form

powder

bp

360 °C (lit.)

mp

147-149 °C (lit.)

solubility

water: soluble 0.00003 g/L at 20 °C

density

1.067 g/mL at 25 °C (lit.)

shipped in

ambient

storage temp.

−20°C

SMILES string

CC(C)CCC[C@@H](C)[C@H]1CC[C@H]2[C@@H]3CC=C4C[C@@H](O)CC[C@]4(C)[C@H]3CC[C@]12C

InChI

1S/C27H46O/c1-18(2)7-6-8-19(3)23-11-12-24-22-10-9-20-17-21(28)13-15-26(20,4)25(22)14-16-27(23,24)5/h9,18-19,21-25,28H,6-8,10-17H2,1-5H3/t19-,21+,22+,23-,24+,25+,26+,27-/m1/s1

InChI key

HVYWMOMLDIMFJA-DPAQBDIFSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Application

Cholesterol was used to prepare immune-stimulating complexes to enhance cellular and humoral responses. It was incorporated in animal feed to study the effect of cholesterol-rich diet.
Standard for chromatography

Biochem/physiol Actions

Cholesterol is a lipid that makes up about 20-25% of the structural components of the cell membranes. It determines the fluidity and permeability of the membrane, making it permeable to water but not to ions and protons. Cholesterol also regulates the functions of the transporters and signaling proteins present on the plasma membrane. The major sites of cholesterol synthesis are small intestine and liver.
Major component of all biological membranes; ~25% of total brain lipid is cholesterol.

Preparation Note

Cholesterol monohydrate yields a clear, colorless to faint yellow solution in hot alcohol. It is also soluble in acetone, dioxane, ethyl acetate, benzene, petroleum ether, oils, fats, and in aqueous solutions of bile salts. Solutions should be protected from light.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jeffrey Buter et al.
Nature chemical biology, 15(9), 889-899 (2019-08-21)
Mycobacterium tuberculosis (Mtb) is the world's most deadly pathogen. Unlike less virulent mycobacteria, Mtb produces 1-tuberculosinyladenosine (1-TbAd), an unusual terpene nucleoside of unknown function. In the present study 1-TbAd has been shown to be a naturally evolved phagolysosome disruptor. 1-TbAd
Effects of 2 or 5 consecutive exercise days on adipocyte area and lipid parameters in Wistar rats
Guerra RL et al
Lipids in Health and Disease, 6, doi: 10-doi: 10 (2007)
Effects of probiotic bacteria, isoflavones and simvastatin on lipid profile and atherosclerosis in cholesterol-fed rabbits: a randomized double-blind study
Cavallini DC et al
Lipids in Health and Disease, 8, doi: 10-doi: 10 (2009)
Anja Helgeby et al.
Journal of immunology (Baltimore, Md. : 1950), 176(6), 3697-3706 (2006-03-07)
The cholera toxin A1 (CTA1)-DD/QuilA-containing, immune-stimulating complex (ISCOM) vector is a rationally designed mucosal adjuvant that greatly potentiates humoral and cellular immune responses. It was developed to incorporate the distinctive properties of either adjuvant alone in a combination that exerted
M A Nurseitova et al.
Journal of dairy science, 102(9), 7723-7733 (2019-07-01)
Milk fat adulteration is a common issue in Central Asia. To assess the current situation in the commercial milk market, 17 milk samples were checked for fatty acid (FA) and sterol profiles to detect potential adulteration using multivariate analysis. Analysis

Articles

Cholesterol biosynthesis starts in the hepatic endoplasmic reticulum with acetyl-CoA, yielding 3-hydroxy-3-methylglutaryl-CoA via HMG-CoA synthase.

Cholesterol biosynthesis starts in the hepatic endoplasmic reticulum with acetyl-CoA, yielding 3-hydroxy-3-methylglutaryl-CoA via HMG-CoA synthase.

Cholesterol biosynthesis starts in the hepatic endoplasmic reticulum with acetyl-CoA, yielding 3-hydroxy-3-methylglutaryl-CoA via HMG-CoA synthase.

Cholesterol biosynthesis starts in the hepatic endoplasmic reticulum with acetyl-CoA, yielding 3-hydroxy-3-methylglutaryl-CoA via HMG-CoA synthase.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service