Skip to Content
Merck
All Photos(1)

Key Documents

P0090000

Palmitic acid

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

1-Pentadecanecarboxylic acid, C16:0, Cetylic acid, Hexadecanoic acid, NSC 5030, PamOH

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)14COOH
CAS Number:
Molecular Weight:
256.42
Beilstein:
607489
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

Agency

EP Reference Standard

vapor pressure

10 mmHg ( 210 °C)

API family

palmitic acid

manufacturer/tradename

EDQM

bp

271.5 °C/100 mmHg (lit.)

mp

61-62.5 °C (lit.)

density

0.852 g/mL at 25 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

CCCCCCCCCCCCCCCC(O)=O

InChI

1S/C16H32O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16(17)18/h2-15H2,1H3,(H,17,18)

InChI key

IPCSVZSSVZVIGE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Palmitic acid EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

235.4 °F

Flash Point(C)

113 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B Vessby et al.
Diabetologia, 37(10), 1044-1050 (1994-10-01)
Recent data indicate that peripheral insulin sensitivity may be influenced by dietary fat quality and skeletal muscle phospholipid fatty acid composition. During a health survey of 70-year-old men insulin sensitivity was measured by the euglycaemic hyperinsulinaemic clamp technique and the
Jonas Sieber et al.
The American journal of pathology, 183(3), 735-744 (2013-07-23)
Type 2 diabetes mellitus is characterized by dyslipidemia with elevated free fatty acids (FFAs). Loss of podocytes is a hallmark of diabetic nephropathy, and podocytes are highly susceptible to saturated FFAs but not to protective, monounsaturated FFAs. We report that
Siri Malmgren et al.
The Journal of biological chemistry, 288(17), 11973-11987 (2013-03-12)
Lipotoxicity is a presumed pathogenetic process whereby elevated circulating and stored lipids in type 2 diabetes cause pancreatic β-cell failure. To resolve the underlying molecular mechanisms, we exposed clonal INS-1 832/13 β-cells to palmitate for 48 h. We observed elevated
Mathieu Blanc et al.
Biochemical Society transactions, 41(1), 84-88 (2013-01-30)
S-Palmitoylation, the only reversible post-translational lipid modification, confers unique biochemical and functional properties to proteins. Although it has long been known that viral proteins are palmitoylated, recent studies reveal that this modification plays a critical role for pathogens of all
Sanja Blaskovic et al.
The FEBS journal, 280(12), 2766-2774 (2013-04-05)
S-palmitoylation is post-translational modification, which consists in the addition of a C16 acyl chain to cytosolic cysteines and which is unique amongst lipid modifications in that it is reversible. It can thus, like phosphorylation or ubiquitination, act as a switch.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service