Skip to Content

O3380

Oleyl oleate

≥99%

Synonym(s):

Oleyl oleate, Oleic acid oleyl ester

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C36H68O2
CAS Number:
Molecular Weight:
532.92
NACRES:
NA.25
PubChem Substance ID:
UNSPSC Code:
12352211
EC Number:
222-980-0
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥99%

form

liquid

functional group

ester, oleic acid

lipid type

unsaturated FAs

shipped in

ambient

storage temp.

−20°C

SMILES string

CCCCCCCC\C=C/CCCCCCCCOC(=O)CCCCCCC\C=C/CCCCCCCC

InChI

1S/C36H68O2/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-33-35-38-36(37)34-32-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2/h17-20H,3-16,21-35H2,1-2H3/b19-17-,20-18-

InChI key

BARWIPMJPCRCTP-CLFAGFIQSA-N

Application


  • Group 13 Lewis acid catalyzed synthesis of metal oxide nanocrystals via hydroxide transmetallation.: This research utilizes oleyl oleate as a solvent to facilitate the synthesis of metal oxide nanocrystals, showcasing its effectiveness in nanoparticle production and potential applications in various electronic and optical devices (Gibson et al., 2021).

  • High-level accumulation of oleyl oleate in plant seed oil by abundant supply of oleic acid substrates to efficient wax ester synthesis enzymes.: This article details the genetic engineering approaches to enhance oleyl oleate content in plant oils, aiming at industrial applications such as biofuels and biolubricants (Yu et al., 2018).



Still not finding the right product?

Explore all of our products under Oleyl oleate


Storage Class

10 - Combustible liquids

wgk

WGK 3

flash_point_f

452.3 °F

flash_point_c

233.5 °C



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library