Merck
  • Clopidogrel bioactivation and risk of bleeding in patients cotreated with angiotensin-converting enzyme inhibitors after myocardial infarction: a proof-of-concept study.
Clopidogrel bioactivation and risk of bleeding in patients cotreated with angiotensin-converting enzyme inhibitors after myocardial infarction: a proof-of-concept study.
Clinical pharmacology and therapeutics (2014-09-16)
K E Kristensen, H-J Zhu, X Wang, G H Gislason, C Torp-Pedersen, H B Rasmussen, J S Markowitz, P R Hansen
ABSTRACT
Clopidogrel is an oral antiplatelet prodrug, the majority of which is hydrolyzed to an inactive metabolite by hepatic carboxylesterase 1 (CES1). Most angiotensin-converting enzyme inhibitors (ACEIs) are also metabolized by this enzyme. We examined the effects of ACEIs on clopidogrel bioactivation in vitro and linked the results with a pharmacoepidemiological study. In vitro, ACEIs inhibited CES1-mediated hydrolysis of a model substrate, and trandolapril and enalapril increased formation of clopidogrel active metabolite. In 70,934 patients with myocardial infarction, hazard ratios for clinically significant bleeding in ACEI-treated patients cotreated with or without clopidogrel were 1.10 (95% confidence interval (CI): 0.97-1.25, P = 0.124) and 0.90 (95% CI: 0.81-0.99, P = 0.025), respectively, as compared with patients who did not receive ACEIs. This difference was statistically significant (P = 0.002). We conclude that cotreatment with selected ACEIs and clopidogrel may increase the risk of bleeding. Combination of in vitro and pharmacoepidemiological studies may be a useful paradigm for assessment of drug-drug interactions.
Sigma-Aldrich
Esterase from Bacillus stearothermophilus, ≥0.2 U/mg
Sigma-Aldrich
2-Bromo-3′-methoxyacetophenone, 98%
Sigma-Aldrich
Esterase Pseudomonas fluorescens, recombinant from E. coli, ≥4 U/mg
Sigma-Aldrich
Perindopril erbumine
Sigma-Aldrich
Esterase from Bacillus stearothermophilus, recombinant, expressed in E. coli, ≥4.0 U/mg
Supelco
Enalapril Maleate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ramipril, ≥98% (HPLC)
USP
Enalapril maleate, United States Pharmacopeia (USP) Reference Standard
USP
Ramipril, United States Pharmacopeia (USP) Reference Standard
Enalapril maleate, European Pharmacopoeia (EP) Reference Standard
Supelco
Ramipril, Pharmaceutical Secondary Standard; Certified Reference Material
Perindopril tert-butylamine, European Pharmacopoeia (EP) Reference Standard
Perindopril for stereochemical purity, European Pharmacopoeia (EP) Reference Standard
Ramipril, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
4-Nitrophenol solution, 10 mM
Sigma-Aldrich
Esterase from porcine liver, lyophilized, powder, slightly beige, ≥50 U/mg
Sigma-Aldrich
4-Nitrophenol, ReagentPlus®, ≥99%
Sigma-Aldrich
Esterase from porcine liver, lyophilized powder, ≥15 units/mg solid
Supelco
Ticlopidine hydrochloride, analytical standard, for drug analysis
Sigma-Aldrich
Esterase from rabbit liver, lyophilized powder, ≥30 units/mg protein
Sigma-Aldrich
4-Nitrophenol, spectrophotometric grade
Sigma-Aldrich
Esterase from porcine liver, ammonium sulfate suspension, ≥150 units/mg protein (biuret)
Ticlopidine hydrochloride, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Esterase Isoenzyme 1 porcine liver, recombinant, recombinant, expressed in E. coli, ≥30.0 U/g
Sigma-Aldrich
Moexipril hydrochloride, powder, ≥98% (HPLC)
Supelco
Acetaminophen Related Compound F, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Esterase from Bacillus subtilis, recombinant, expressed in E. coli, ≥10 U/mg
Sigma-Aldrich
Fosinopril sodium, ≥98% (HPLC), powder
USP
Fosinopril sodium, United States Pharmacopeia (USP) Reference Standard
Supelco
4-Nitrophenol, PESTANAL®, analytical standard