Skip to Content
Merck
All Photos(2)

Documents

1048

Sigma-Aldrich

4-Nitrophenol

spectrophotometric grade

Synonym(s):

p-Nitrophenol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
O2NC6H4OH
CAS Number:
Molecular Weight:
139.11
Beilstein:
1281877
EC Number:
MDL number:
UNSPSC Code:
12352102
eCl@ss:
39032018
PubChem Substance ID:
NACRES:
NA.21

grade

spectrophotometric grade

Quality Level

vapor pressure

0.6 mmHg ( 120 °C)

form

powder

autoignition temp.

541 °F

technique(s)

UV/Vis spectroscopy: suitable

bp

279 °C (lit.)

mp

110-115 °C (lit.)

solubility

ethanol: 95%, clear, dark yellow (100 mg/mL)

SMILES string

O=N(C1=CC=C(O)C=C1)=O

InChI

1S/C6H5NO3/c8-6-3-1-5(2-4-6)7(9)10/h1-4,8H

InChI key

BTJIUGUIPKRLHP-UHFFFAOYSA-N

Gene Information

human ... UGT1A4(54657)

Looking for similar products? Visit Product Comparison Guide

Application


  • Spectroscopic monitoring of polyurethane-based nanocomposite as a potential catalyst for the reduction of dyes.: This research explores the use of a polyurethane-based nanocomposite incorporating 4-Nitrophenol for spectroscopic monitoring and catalysis. The study demonstrates its effectiveness in reducing dye concentrations, providing a valuable tool for environmental cleanup and chemical processing industries (Alam K et al., 2024).

  • Silica@poly(chitosan-N-isopropylacrylamide-methacrylic acid) microgels: Extraction of palladium (II) ions and in situ formation of palladium nanoparticles for pollutant reduction.: Discusses the synthesis and application of innovative microgels that utilize 4-Nitrophenol in processes for extracting palladium ions and reducing pollutants. This highlights its utility in heavy metal remediation and nanoparticle synthesis, relevant for environmental and materials science (Alzahrani AYA et al., 2024).

  • Reaction mechanisms of chlorinated disinfection byproducts formed from nitrophenol compounds with different structures during chlor(am)ination and UV/post-chlor(am)ination.: Examines how 4-Nitrophenol and similar compounds react under different water treatment processes, providing insights into the formation and control of disinfection byproducts. This is crucial for improving water treatment safety and effectiveness (Prasad Singh R et al., 2024).

  • Comprehensive Studies of Adsorption Equilibrium and Kinetics for Selected Aromatic Organic Compounds on Activated Carbon.: This study provides a detailed analysis of 4-Nitrophenol adsorption on activated carbon, offering valuable data on its behavior in water treatment systems. The findings are critical for designing more efficient adsorption processes to remove contaminants from water (Marczewski AW et al., 2024).


Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - STOT RE 2 - STOT RE 2 Oral

Target Organs

Kidney,Liver

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

336.2 °F

Flash Point(C)

169 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Mitrofanova et al.
Nature communications, 10(1), 2692-2692 (2019-06-21)
Sphingomyelin phosphodiesterase acid-like 3b (SMPDL3b) is a lipid raft enzyme that regulates plasma membrane (PM) fluidity. Here we report that SMPDL3b excess, as observed in podocytes in diabetic kidney disease (DKD), impairs insulin receptor isoform B-dependent pro-survival insulin signaling by
Ali Mehdinia et al.
Journal of chromatography. A, 1283, 82-88 (2013-03-08)
In this study, an analytical procedure for the selective extraction and detection of 4-nitrophenol (4-NP) was investigated by using of molecularly imprinted polymer on the surface of magnetic nanoparticles (MNPs). The magnetic nanoparticles were modified by tetraethyl orthosilicate (TEOS) and
Alex V Birk et al.
The Journal of laboratory and clinical medicine, 140(3), 166-175 (2002-09-25)
Extracellular adenosine triphosphate (ATP) regulates platelet reactivity by way of direct action on platelet purinergic receptors or by hydrolysis to adenosine diphosphate (ADP). Subsequent metabolism of ATP and ADP to adenosine monophosphate (AMP) and adenosine inhibits platelet aggregation. Endothelial cell
Ipsita K Sen et al.
Carbohydrate polymers, 91(2), 518-528 (2012-11-06)
Gold nanoparticles were synthesized by reducing chloroauric acid with a glucan, isolated from an edible mushroom Pleurotus florida, cultivar Assam Florida. Here, glucan acts as reducing as well as stabilizing agent. The synthesized gold nanoparticles were characterized by UV-visible spectroscopy
Siew Hong Lam et al.
Biochimica et biophysica acta, 1830(10), 4778-4789 (2013-06-25)
4-Nitrophenol (4-NP) is a prioritized environmental pollutant and its toxicity has been investigated using zebrafish, advocated as an alternative toxicological model. However, molecular information of 4-NP induced hepatotoxicity is still limited. This study aimed to obtain molecular insights into 4-NP-induced

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service