Skip to Content
Merck
All Photos(1)

Key Documents

F1308

Sigma-Aldrich

Fosinopril sodium

≥98% (HPLC), powder

Synonym(s):

(2S,4S)-4-Cyclohexyl-1-(2-{[2-methyl-1-(propanoyloxy)propoxy](4-phenylbutyl)phosphoryl}acetyl)pyrrolidine-2-carboxylic acid, Acecor, Eosinopril, Fosinopril, Monopril, SQ 28555, Secorvas, Staril

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H45NO7P · Na
CAS Number:
Molecular Weight:
585.64
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to off-white

solubility

H2O: >20 mg/mL

originator

Bristol-Myers Squibb

storage temp.

−20°C

SMILES string

O=C([O-])[C@H]1N(C(CP(CCCCC2=CC=CC=C2)(O[C@@H](C(C)C)OC(CC)=O)=O)=O)C[C@H](C3CCCCC3)C1.[Na+]

InChI

1S/C30H46NO7P.Na/c1-4-28(33)37-30(22(2)3)38-39(36,18-12-11-15-23-13-7-5-8-14-23)21-27(32)31-20-25(19-26(31)29(34)35)24-16-9-6-10-17-24;/h5,7-8,13-14,22,24-26,30H,4,6,9-12,15-21H2,1-3H3,(H,34,35);/q;+1/p-1/t25-,26+,30+,39?;/m1./s1

InChI key

TVTJZMHAIQQZTL-TXDYNIFHSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Fosinopril lowers blood pressure and functions as an antihypertensive agent. It is an ester prodrug. Fosinopril is metabolised by the liver and excreted from the body by liver and kidney. It is used to treat heart failure.

Biochem/physiol Actions

Fosinopril is an angiotensin converting enzyme (ACE) inhibitor.
Fosinopril is an angiotensin converting enzyme (ACE) inhibitor. Fosinopril is also known to inhibit the activity of the peptide transporter PEPT2.

Features and Benefits

This compound is a featured product for ADME Tox research. Click here to discover more featured ADME Tox products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound was developed by Bristol-Myers Squibb. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Giovannini et al.
The journal of nutrition, health & aging, 14(6), 457-460 (2010-07-10)
The present study evaluates the effects of a 6-month treatment with an ACE-inhibitor (ie, fosinopril) on serum concentrations of total IGF-1 and IGF binding protein (IGFBP)-3 in older adults at high risk for cardiovascular disease. Data are from the Trial
Dinesh Shrikrishna et al.
Chest, 146(4), 932-940 (2014-02-22)
Skeletal muscle impairment is a recognized complication of COPD, predicting mortality in severe disease. Increasing evidence implicates the renin-angiotensin system in control of muscle phenotype. We hypothesized that angiotensin-converting enzyme (ACE) inhibition would improve quadriceps function and exercise performance in
Disposition of fosinopril sodium in healthy subjects
Singhv SM, et al.
British Journal of Clinical Pharmacology, 25(1), 9-15 (1988)
Patrick Rossignol et al.
Hypertension (Dallas, Tex. : 1979), 60(2), 339-346 (2012-07-11)
Optimal blood pressure (BP) targets are still controversial in end-stage renal disease. Recent data have highlighted shortcomings of the usual BP hypothesis in other patient populations and emphasized the importance of visit-to-visit variability of BP in predicting cardiovascular events. The
Ya-chen Zhang et al.
Cell biochemistry and biophysics, 64(3), 205-211 (2012-06-26)
Fosinopril, an angiotensin-converting enzyme inhibitor, is known to attenuate cardiomyopathy induced by doxorubicin (DOX); however, the mechanisms of this cardioprotection are not fully elucidated yet. In the present study, experimental cardiomyopathy was induced in rats by administration of DOX with

Articles

We offer many products related to angiotensin receptors for your research needs.

We offer many products related to angiotensin receptors for your research needs.

We offer many products related to angiotensin receptors for your research needs.

We offer many products related to angiotensin receptors for your research needs.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service