Skip to Content
Merck
All Photos(1)

Key Documents

P0094

Sigma-Aldrich

Perindopril erbumine

Synonym(s):

(2S,3aS,7aS)-1-[(2S)-2-[[(1S)-1-(Ethoxycarbonyl)butyl]amino]-1-oxopropyl]octahydro-1H-indole-2-carboxylic acid tert-butylamine salt, Perindopril tert-butylamine, S-9490

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H32N2O5 · C4H11N
CAS Number:
Molecular Weight:
441.60
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

Quality Level

form

powder

solubility

H2O: 10 mg/mL, clear

originator

Xoma

storage temp.

2-8°C

SMILES string

CC(C)(C)N.CCC[C@H](N[C@@H](C)C(=O)N1[C@H]2CCCC[C@H]2C[C@H]1C(O)=O)C(=O)OCC

InChI

1S/C19H32N2O5.C4H11N/c1-4-8-14(19(25)26-5-2)20-12(3)17(22)21-15-10-7-6-9-13(15)11-16(21)18(23)24;1-4(2,3)5/h12-16,20H,4-11H2,1-3H3,(H,23,24);5H2,1-3H3/t12-,13-,14-,15-,16-;/m0./s1

InChI key

IYNMDWMQHSMDDE-MHXJNQAMSA-N

Gene Information

human ... ACE(1636)

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Perindopril erbumine is an angiotensin converting enzyme (ACE) inhibitor; antihypertensive.
Perindopril erbumine is an angiotensin converting enzyme (ACE) inhibitor; antihypertensive; becomes hydrolyzed in vivo to the active diacid metabolite; unlike the other ACE inhibitors, inhibits tumor growth in hepatocellular carcinoma cells due to suppression of VEGF levels; also suppresses angiotensin II production in vitro. Long-term therapy with this agent has a beneficial effect on the cerebral circulation by improving cerebral perfusion reserve in patients with previous minor stroke.

Features and Benefits

This compound is featured on the Angiotensin Receptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.
This compound was developed by Xoma. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Chris Tikellis et al.
Clinical science (London, England : 1979), 124(10), 617-626 (2012-12-12)
It is recommended that individuals with diabetes restrict their dietary sodium intake. However, although salt intake is correlated with BP (blood pressure), it also partly determines the activation state of the RAAS (renin-angiotensin-aldosterone system), a key mediator of diabetes-associated atherosclerosis.
Kate T Murphy et al.
International journal of cancer, 133(5), 1234-1246 (2013-02-26)
Cancer cachexia describes the progressive skeletal muscle wasting and weakness associated with many cancers. Cachexia reduces mobility and quality of life and accounts for 20-30% of all cancer-related deaths. Activation of the renin-angiotensin system causes skeletal muscle wasting and weakness.
Linda Battes et al.
The American journal of cardiology, 112(1), 27-33 (2013-04-06)
Appropriate risk stratification of patients with established, stable coronary artery disease could contribute to the prevention of recurrent cardiovascular events. The purpose of the present study was to develop and validate risk prediction models for various cardiovascular end points in
Omar Mukhtar et al.
British journal of clinical pharmacology, 75(4), 951-954 (2012-08-18)
Early trials in the field of hypertension focused on adults in their fifties and sixties. However, with the passage of time, a progressive effort has been made to expand the evidence base for treatment in older adults. 2008 saw publication
Christopher J Bulpitt et al.
Hypertension (Dallas, Tex. : 1979), 61(1), 89-94 (2012-11-23)
White coat hypertension is considered to be a benign condition that does not require antihypertensive treatment. Ambulatory blood pressure (ABP) was measured in 284 participants in the Hypertension in the Very Elderly Trial (HYVET), a double-blind randomized trial of indapamide

Articles

We offer many products related to angiotensin receptors for your research needs.

We offer many products related to angiotensin receptors for your research needs.

We offer many products related to angiotensin receptors for your research needs.

We offer many products related to angiotensin receptors for your research needs.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service