Skip to Content
Merck
All Photos(1)

Key Documents

T6654

Supelco

Ticlopidine hydrochloride

analytical standard, for drug analysis

Synonym(s):

5-(o-Chlorobenzyl)-4,5,6,7-tetrahydrothieno[3,2-c]pyridine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H14ClNS · HCl
CAS Number:
Molecular Weight:
300.25
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard, for drug analysis

Quality Level

Assay

≥99%

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)
veterinary

format

neat

SMILES string

Cl.Clc1ccccc1CN2CCc3sccc3C2

InChI

1S/C14H14ClNS.ClH/c15-13-4-2-1-3-11(13)9-16-7-5-14-12(10-16)6-8-17-14;/h1-4,6,8H,5,7,9-10H2;1H

InChI key

MTKNGOHFNXIVOS-UHFFFAOYSA-N

Gene Information

human ... P2RY12(64805)

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Ticlopidine hydrochloride has been used as an analytical standard to investigate the developmental toxicity and teratogenic potential of ticlopidine in Xenopus laevis embryos and human endothelial cells using a frog embryo teratogenesis assay-Xenopus (FETAX) and blood and lymph vessel formation assay.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Characterization of ticlopidine-induced developmental and teratogenic defects in Xenopus embryos and human endothelial cells.
Park SM, et al.
Chemico-Biological Interactions, 240, 172-178 (2015)
Morten Lamberts et al.
Circulation, 129(15), 1577-1585 (2014-01-29)
The optimal long-term antithrombotic treatment of patients with coexisting atrial fibrillation and stable coronary artery disease is unresolved, and commonly, a single antiplatelet agent is added to oral anticoagulation. We investigated the effectiveness and safety of adding antiplatelet therapy to
Nohra Chalouhi et al.
Stroke, 45(1), 54-58 (2013-11-21)
Flow diverters are currently indicated for treatment of large and complex intracranial aneurysms. The purpose of this study was to determine whether the indications of flow diversion can be safely extended to unruptured, small, saccular aneurysms (<10 mm) of the
Julie A Johnson et al.
Pharmacological reviews, 65(3), 987-1009 (2013-05-21)
The past decade has seen tremendous advances in our understanding of the genetic factors influencing response to a variety of drugs, including those targeted at treatment of cardiovascular diseases. In the case of clopidogrel, warfarin, and statins, the literature has
Stephen D Wiviott et al.
Lancet (London, England), 382(9892), 605-613 (2013-08-21)
Treatment with prasugrel and aspirin improves outcomes compared with clopidogrel and aspirin for patients with acute coronary syndrome who have had angiography and percutaneous coronary intervention; however, no clear benefit has been shown for patients managed first with drugs only.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service