Skip to Content
Merck

FG0465

IMDM

With stable-glutamine and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture

Synonym(s):

Cell culture medium, Iscove′s MDM, Iscove’s Modified Dulbecco’s Medium, Modified Dulbecco′s medium

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352207
NACRES:
NA.71
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Iscove′s Modified Dulbecco′s Medium, With L-alanyl-L-glutamine and sodium bicarbonate, sterile-filtered, liquid, suitable for cell culture

Quality Segment

sterility

sterile-filtered, sterile

product line

AQmedia

form

liquid

technique(s)

cell culture | mammalian: suitable

impurities

Endotoxin, tested

components

stable glutamine: yes
NaHCO3: yes
glucose: yes
phenol red: yes

shipped in

ambient

storage temp.

2-8°C

General description

This IMDM differs from the original formula, because it is supplemented with L-alanyl-L-glutamine dipeptide. Glutamine is notoriously unstable in cell culture. This dipeptide provides a more stable form of glutamine for use in cell cultures.

Application

Guilbert and Iscove demonstrated that precursor cells of erythrocytes and macrophages could be cultured in a totally defined serum free medium supplemented with albumin, transferrin, lecithin, and selenium. This medium is a modification of Dulbecco¢s Modified Eagle¢s Medium (DME) containing selenium, additional amino acids and vitamins, sodium pyruvate, HEPES buffer, and potassium nitrate instead of ferric nitrate. Further studies demonstrated that Iscove¢s Medium would support murine B lymphocytes, hemopoietic tissue from bone marrow, B cells stimulated with lipopolysaccharide, T lymphocytes, and a variety of hybrid cells.

Legal Information

AQmedia is a trademark of Sigma-Aldrich Co. LLC


Still not finding the right product?


Storage Class

10 - Combustible liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library