Skip to Content
Merck

SEGPU0538

Stericup® E-GP Funnel-less Sterile Vacuum Filtration System

greener alternative

pore size 0.22 μm, polyethersulfone membrane, filter capacity 500 mL, 38 mm (1 1/2 in.) thread, pack of 12 ea

Synonym(s):

Stericup® E-GP Eco-friendly Filter Unit

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NB.24
UNSPSC Code:
41104914
Pore size:
0.22 μm pore size
Filter capacity:
500 mL
Packaging:
pack of 12 ea
Material:
polyethersulfone membrane, polystyrene bottle, polystyrene funnel(s), polystyrene housing
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

material

polyethersulfone membrane, polystyrene bottle, polystyrene funnel(s), polystyrene housing

product line

Stericup®

feature

hydrophilic

packaging

pack of 12 ea

greener alternative product characteristics

Waste Prevention
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

parameter

max. temp. at 45 °C

description

2.4 mL (hold-up volume)

technique(s)

sterile filtration: suitable

H

193 mm

diam.

9.3 cm

filter capacity

500 mL

filter diam.

73 mm

filtration area

40 cm2

matrix

Express PLUS

pore size

0.22 μm pore size

fitting

inlet thread(s), 38 mm (1 1/2 in.) thread

greener alternative category

shipped in

ambient

storage temp.

no temp limit

General description

We are committed to bringing you Greener Alternative Products, which belong to one of the four categories of greener alternatives . This product is a Design for Sustainability (DfS) developed product. Click here to view its DfS scorecard.
Work with ease and filter with confidence with Stericup® and Steritop® Filtration Systems.

The Stericup® E and Steritop® E sterile filters are designed as a Greener Alternative for vacuum sterile filtration that significantly reduces plastic and packaging.

This Stericup® E is a 38mm thread collar which uses a 0.22 μm pore size polyethersulfone (PES) membrane filter unit combined with a 500 mL receiver flask and cap for processing and storage. The 38 mm thread directly connects to our media bottles and majority of other standard commercial media bottles.

Use only glass or plastic bottles designed for vacuum applications.

Application

For Research Use Only
Stericup® and Steritop® systems are designed to reduce foaming and protein denaturation while maximizing flow and can be used for sterile vacuum filtration of aqueous solutions such as tissue culture media and biological fluids.

  • Tissue culture media +/- additives
  • Buffers
  • Biological solutions

Features and Benefits

  • The filter collar disconnects from the receiver bottle with just a quarter turn—reducing the likelihood of spillage and making it easier to manipulate on the bench or under the hood.
  • Bold product identification on the filter collar plainly displays membrane characteristics, including composition and pore size, for rapid, application-appropriate product selection.
  • Click seal cap is flanged and modified to enable a firm grip in wet or dry conditions—and a tactile stop confirms secure closure. Worry less about the possibility of spilling or contamination of sterilized contents.
  • Frosted, ample writing surface on the receiver bottle and lighter cap color facilitate clear labelling to improve legibility and reduce unreadable smears.
  • Click seal cap rests on the side to avoid risk of contamination.
  • Receiver bottles are stackable to save storage space.

Legal Information

EXPRESS PLUS is a trademark of Merck KGaA, Darmstadt, Germany
STERITOP is a registered trademark of Merck KGaA, Darmstadt, Germany
Stericup is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?




Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library





Global Trade Item Number

SKUGTIN
SEGPU053804061841074236