Skip to Content
Merck

S2GPU05RE

Millipore® Stericup® Quick Release Vacuum Filtration System

pore size 0.22 μm, polyethersulfone membrane, filter capacity 500 mL, pack of 12 ea

Synonym(s):

SCGPU05RE

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41104914
NACRES:
NB.24
eCl@ss:
32031623
Pore size:
0.22 μm pore size
Filter capacity:
500 mL
Packaging:
pack of 12 ea
Material:
polyethersulfone membrane, polystyrene bottle, polystyrene funnel(s), polystyrene housing
Sterility:
sterile
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

material

polyethersulfone membrane, polystyrene bottle, polystyrene funnel(s), polystyrene housing

sterility

sterile

product line

Stericup®

feature

hydrophilic

packaging

pack of 12 ea

parameter

45 °C max. temp.

description

2.4 mL (hold-up volume)

technique(s)

sterile filtration: suitable

H

263 mm

diam.

12.5 cm

filter capacity

500 mL

filter diam.

73 mm

filtration area

40 cm2

matrix

Express PLUS

pore size

0.22 μm pore size

fitting

inlet funnel, outlet flask (Receiver)

shipped in

ambient

storage temp.

15-25°C

General description

Stericup Quick Release-GP sterile vacuum filtration system combines a 500 mL Steritop bottle top filter unit, which uses a 0.22 µm pore size fast flow PES Express PLUS membrane, with a 500 mL receiver flask for sterilizing tissue culture media, protein & DNA.

Application

For Research Use Only
Stericup® and Steritop® systems are designed to reduce foaming and protein denaturation while maximizing flow and can be used for sterile vacuum filtration of aqueous solutions such as tissue culture media and biological fluids.

  • Tissue culture media +/- additives
  • Buffers
  • Biological solutions

Features and Benefits

  • The Quick Release filter funnel disconnects from the receiver bottle with just a quarter turn—reducing the likelihood of spillage and making it easier to manipulate on the bench or under the hood.
  • Bold product identification on the Steritop® filter funnel plainly displays membrane characteristics, including composition and pore size, for rapid, application-appropriate product selection.
  • Click seal cap is flanged and modified to enable a firm grip in wet or dry conditions—and a tactile stop confirms secure closure. Worry less about the possibility of spilling or contamination of sterilized contents.
  • Frosted, ample writing surface on the receiver bottle and lighter cap color facilitate clear labelling to improve legibility and reduce unreadable smears.
  • Click seal cap rests on the side to avoid risk of contamination.
  • Receiver bottles are stackable to save storage space.

Legal Information

EXPRESS PLUS is a trademark of Merck KGaA, Darmstadt, Germany
Millipore is a registered trademark of Merck KGaA, Darmstadt, Germany
Stericup is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?




Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library





Global Trade Item Number

SKUGTIN
S2GPU05RE04054839116490